  • Search Results
  • Cation ion specifically induces a conformational change in trans-dehydroandrosterone - a solid-state NMR study.
Cation ion specifically induces a conformational change in trans-dehydroandrosterone - a solid-state NMR study.
Steroids (2015-02-01)
Darong Wang, Meiman Chen, Rong-Jie Chein, Wei-Min Ching, Chen-Hsiung Hung, Der-Lii M Tzou
ABSTRACT
In this work, we demonstrated that calcium (Ca(+2)) is able to induce a conformational change in trans-dehydroandrosterone (DHEA). To this respect, solid-state NMR spectroscopy was applied to a series of DHEA molecules that were incubated with Ca(+2) under different concentrations. The high-resolution (13)C NMR spectra of the DHEA/Ca(+2) mixtures exhibited two distinct sets of signals; one was attributed to DHEA in the free form, and the second set was due to the DHEA/Ca(+2) complex. Based on chemical shift isotropy and anisotropy analyses, we postulated that Ca(+2) might have associated with the oxygen attached to C17 via a lone-pair of electrons, which induced a conformational change in DHEA. Apart from Ca(+2), we also incubated DHEA with magnesium (Mg(+2)) to determine whether Mg(+2) was able to interact with DHEA in a similar manner to Ca(+2). We found that Mg(+2) was able to induce a conformational change in DHEA deviated from that of Ca(+2). These solid-state NMR observations indicate that DHEA is able to interact with cations, such as Mg(+2) and Ca(+2), with specificity.
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Allyl methyl sulfone, 96%