  • Search Results
  • Effect of different calcium sources on the bioactive compounds stability of extruded and nixtamalized blue maize flours.
Effect of different calcium sources on the bioactive compounds stability of extruded and nixtamalized blue maize flours.
Journal of food science and technology (2015-04-22)
Miguel Ángel Sánchez-Madrigal, Armando Quintero-Ramos, Fernando Martínez-Bustos, Carmen O Meléndez-Pizarro, Martha G Ruiz-Gutiérrez, Alejandro Camacho-Dávila, Patricia Isabel Torres-Chávez, Benjamín Ramírez-Wong
ABSTRACT
The stability of antioxidants in extruded and nixtamalized blue maize flours with calcium hydroxide [Ca(OH)2] and calcium lactate [C6H10O6Ca] were evaluated. Extruded blue maize flours batches were obtained by mixing blue maize flours separately with different Ca(OH)2 (0.1, 0.2 and 0.3 %) and C6H10O6Ca (0.3, 0.6 and 0.9 %) concentrations respectively and extruded to obtain the extruded flours. For nixtamalized flours, the maize grains were cooked at 1 % Ca(OH)2 and 2.95 % C6H10O6Ca concentrations respectively. Color, antioxidant activity, total phenolics, total anthocyanins and individual anthocyanins, contents were analyzed. Color, antioxidant activity, anthocyanins contents and total phenolics decreased as the calcium hydroxide concentration increased. In contrast, increasing the calcium lactate concentration on the extruded flours had the opposite effect. The extrusion process retained 57-47 %, 72-62 % and 79-65 % of the anthocyanins content, total phenolic content and antioxidant activity, respectively. These retention rates were higher than those of the nixtamalized flours using the same calcium sources. Cyanidin-3-glucoside and pelargonidin-3-glucoside were identified in the maize kernel and flours. Cyanidin-3-glucoside concentration was increased by both extrusion and nixtamalization processed with either of the two calcium sources. In contrast, pelargonidin-3-glucoside concentration decreased by both processes. Other anthocyanins were observed, but they were not identified.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Supelco
Gallic acid, certified reference material, TraceCERT®
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol solution, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, BioReagent, for molecular biology
Sigma-Aldrich
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material