  • Search Results
  • Changes in the distribution of type II transmembrane serine protease, TMPRSS2 and in paracellular permeability in IPEC-J2 cells exposed to oxidative stress.
Changes in the distribution of type II transmembrane serine protease, TMPRSS2 and in paracellular permeability in IPEC-J2 cells exposed to oxidative stress.
Inflammation (2014-08-07)
Erzsebet Paszti-Gere, Reka Fanni Barna, Csaba Kovago, Ipoly Szauder, Gabriella Ujhelyi, Csaba Jakab, Nóra Meggyesházi, Andras Szekacs
ABSTRACT
The effect of oxidative stress on barrier integrity and localization of transmembrane serine proteinase 2 (TMPRSS2) were studied using porcine epithelial IPEC-J2 cells on membrane inserts. Increased paracellular permeability of FITC-dextran 4 kDa (fluorescence intensity 43,508 ± 2,391 versus 3,550 ± 759) and that of gentamicin (3.41 ± 0.06 % increase to controls) were measured parallel with the reduced transepithelial electrical resistance (23.3 ± 4.06 % decrease) of cell layers 6 h after 1 h 1 mM H2O2 treatment. The immunohistochemical localization of adherens junctional β-catenin was not affected by reactive oxygen species (ROS) up to 4 mM H2O2. Peroxide-triggered enhanced paracellular permeability of IPEC-J2 cell layer was accompanied by predominantly cytoplasmic occurrence of TMPRSS2 embedded in cell membrane under physiological conditions. These results support that ROS can influence paracellular gate opening via multifaceted mode of action without involvement of β-catenin redistribution in adherens junction. Altered distribution pattern of TMPRSS2 and relocalized transmembrane serine protease activity may contribute to weakening of epithelial barrier integrity under acute oxidative stress.
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
SAFC
Glycine
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Supelco
Methanol, analytical standard