  • Search Results
  • Application of nano-sized multi-template imprinted polymer for simultaneous extraction of polycyclic aromatic hydrocarbon metabolites in urine samples followed by ultra-high performance liquid chromatographic analysis.
Application of nano-sized multi-template imprinted polymer for simultaneous extraction of polycyclic aromatic hydrocarbon metabolites in urine samples followed by ultra-high performance liquid chromatographic analysis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-11)
Abhishek Chauhan, Tejasvi Bhatia, Amarnath Singh, Prem Narain Saxena, C Kesavchandran, Mohana Krishna Reddy Mudiam
ABSTRACT
Nano-sized molecularly imprinted polymer (nMIP) was synthesized through precipitation polymerization method and used it as solid phase extraction (SPE) sorbent for the selective and simultaneous extraction of hydroxylated metabolites of polycyclic aromatic hydrocarbons (PAHs) from urine followed by ultra-high performance liquid chromatography (UHPLC) analysis coupled with the fluorescent detector (FLD). Multi-template imprinting approach was used in the synthesis of nMIP by taking 1-naphthol, 9-phenanthrol and 9-hydroxyfluorene as templates, methacrylic acid (MAA) as a monomer, ethyleneglycoldimethacrylate (EGDMA) as a crosslinker and AIBN as an initiator. The synthesized nMIP exhibit the highest degree of binding affinity in comparison to non-imprinted polymer (NIP), and its binding affinity found to be in the range of 50-90% for all five metabolites tested. Method exhibits good linearity over a range of concentrations of metabolites with a R(2) value ranges from 0.9789 to 0.9921. Limit of detection (LOD) and limit of quantitation (LOQ) in urine samples were found to be in the range of 0.33-2.6 and 0.99-8ngmL(-1), respectively. Precision study shows that intra and inter-day precision was found to be less than 10%. The application of nMIP as SPE sorbent offers an effective and selective affinity towards the PAH metabolite's and found to be an alternative to the conventional sorbent for PAH metabolite's extraction from the biological samples. The developed nMIP offers wide advantages like simultaneous determination of PAH metabolites with improved sensitivity and found to be cost-effective for the routine analysis.
Sigma-Aldrich
Acetonitrile solution, contains 0.035 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
1-Naphthol, ReagentPlus®, ≥99%
Sigma-Aldrich
1-Naphthol, BioXtra, ≥99%
Sigma-Aldrich
1-Naphthol, puriss. p.a., reag. Ph. Eur., ≥99% (GC)
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), 98%
Sigma-Aldrich
9-Phenanthrol, technical grade
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 40.0% 2-propanol, 0.05% formic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 0.1 % (v/v) formic acid, 5 % (v/v) water, suitable for HPLC
Supelco
1-Naphthol, PESTANAL®, analytical standard
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2,2′-Azobis(2-methylpropionitrile), recrystallized from methanol, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Methyl acetoacetate, ReagentPlus®, 99%
Sigma-Aldrich
4-Vinylpyridine, contains 100 ppm hydroquinone as inhibitor, 95%
Sigma-Aldrich
Methacrylic acid, contains 250 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent