  • Search Results
  • Plasmonic enhancement of dye sensitized solar cells via a tailored size-distribution of chemically functionalized gold nanoparticles.
Plasmonic enhancement of dye sensitized solar cells via a tailored size-distribution of chemically functionalized gold nanoparticles.
PloS one (2014-10-30)
Codrin Andrei, Elena Lestini, Stephen Crosbie, Caoimhe de Frein, Thomas O'Reilly, Dominic Zerulla
ABSTRACT
A substantial and stable increase of the current density Jsc of ruthenium (Ru) dye sensitized solar cells (DSC) of up to 16.18% and of the power efficiency of up to 25.5% is demonstrated in this article via plasmonic enhancement. The key aspect of this work is the use of a tailored bimodal size distribution of functionalized gold nanoparticles (AuNPs) that have been chemically immobilized onto the mesoporous titanium dioxide (TiO2) layer via short, stable dithiodibutyric acid linkers. The size distribution of the AuNPs is a result of theoretical calculations that aimed at the perfection of the absorption characteristics of the complete solar cell system over a wide range of wavelengths. The functionalization of the AuNPs serves to bind them at a close but defined distance to TiO2-particles and additionally to chemically protect them against potential corrosion by the electrolyte. Simulations of near field (enhanced absorption) and far field (scattering) contributions have been used to tailor a complex AuNPs bimodal size distribution that had subsequently demonstrated experimentally a close to optimum improvement of the absorbance over a wide wavelength range (500-675 nm) and therefore an impressive DSC efficiency enhancement. Finally, the modified DSCs are exhibiting pronounced longevity and stable performance as confirmed via long time measurements. In summary, the presented systems show increased performance compared to non plasmonic enhanced cells with otherwise identical composition, and are demonstrating a previously unpublished longevity for iodide electrolyte/AuNPs combinations.
Product Description
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%