  • Search Results
  • Multiplexed microfluidic enzyme assays for simultaneous detection of lipolysis products from adipocytes.
Multiplexed microfluidic enzyme assays for simultaneous detection of lipolysis products from adipocytes.
Analytical and bioanalytical chemistry (2014-06-02)
Colleen E Dugan, William P Cawthorn, Ormond A MacDougald, Robert T Kennedy
ABSTRACT
Microfluidics has enabled new cell biology experiments. Incorporating chemical monitoring of cellular secretion into chips offers the potential to increase information content and utility of such systems. In this work, an integrated, multilayer polydimethylsiloxane microfluidic chip was developed to simultaneously measure fatty acids and glycerol secreted from cultured adipocytes on chip in near real time. Approximately 48,000 adipocytes were loaded into a cell chamber in a reversibly sealed chip. Cells were perfused at 0.75 μL/min. Cell perfusate was split and directed to separate, continuously operating fluorescent enzyme assay channel networks. The fluorescent assay products were detected simultaneously near the outlet of the chip. The fatty acid and glycerol assays had linear dynamic ranges of 150 and 110 μM and limit of detection (LOD) of 6 and 5 μM, respectively. Surface modifications including pretreatment with sodium dodecyl sulfate were utilized to prevent adsorption of fatty acids to the chip surface. Using the chip, basal fatty acid and glycerol concentrations ranged from 0.18 to 0.7 nmol × 10(6) cell(-1) min(-1) and from 0.23 to 0.85 nmol × 10(6) cell(-1) min(-1), respectively. Using valves built into the chip, the perfusion solution was switched to add 20 μM isoproterenol, a β-adrenergic agonist, which stimulates the release of glycerol and fatty acids in adipocytes. This manipulation resulted in a rapid and stable 1.5- to 6.0-fold increase of non-esterified fatty acid (NEFA) and glycerol. The ratio of NEFA:glycerol released increased with adipocyte age. These experiments illustrate the potential for performing multiple real-time assays on cells in culture using microfluidic devices.
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Oleic acid, suitable for cell culture, BioReagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Hexamethyldisilazane, reagent grade, ≥99%
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Hexamethyldisilazane, produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Supelco
N,O-Bis(trimethylsilyl)acetamide, for GC derivatization, LiChropur, ≥98.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Hexamethyldisilazane, for GC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Hexamethyldisilazane, ReagentPlus®, 99.9%