MilliporeSigma
  • The crystal structure of ferritin from Chlorobium tepidum reveals a new conformation of the 4-fold channel for this protein family.
The crystal structure of ferritin from Chlorobium tepidum reveals a new conformation of the 4-fold channel for this protein family.
Biochimie (2014-08-01)
Mauricio Arenas-Salinas, Philip D Townsend, Christian Brito, Valeria Marquez, Vanessa Marabolli, Fernando Gonzalez-Nilo, Cata Matias, Richard K Watt, Juan D López-Castro, José Domínguez-Vera, Ehmke Pohl, Alejandro Yévenes
ABSTRACT
Ferritins are ubiquitous iron-storage proteins found in all kingdoms of life. They share a common architecture made of 24 subunits of five α-helices. The recombinant Chlorobium tepidum ferritin (rCtFtn) is a structurally interesting protein since sequence alignments with other ferritins show that this protein has a significantly extended C-terminus, which possesses 12 histidine residues as well as several aspartate and glutamic acid residues that are potential metal ion binding residues. We show that the macromolecular assembly of rCtFtn exhibits a cage-like hollow shell consisting of 24 monomers that are related by 4-3-2 symmetry; similar to the assembly of other ferritins. In all ferritins of known structure the short fifth α-helix adopts an acute angle with respect to the four-helix bundle. However, the crystal structure of the rCtFtn presented here shows that this helix adopts a new conformation defining a new assembly of the 4-fold channel of rCtFtn. This conformation allows the arrangement of the C-terminal region into the inner cavity of the protein shell. Furthermore, two Fe(III) ions were found in each ferroxidase center of rCtFtn, with an average FeA-FeB distance of 3 Å; corresponding to a diferric μ-oxo/hydroxo species. This is the first ferritin crystal structure with an isolated di-iron center in an iron-storage ferritin. The crystal structure of rCtFtn and the biochemical results presented here, suggests that rCtFtn presents similar biochemical properties reported for other members of this protein family albeit with distinct structural plasticity.
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%
Sigma-Aldrich
Thioglycolic acid solution, ~70 % (w/w) in H2O
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Sodium hydrosulfite, ≥82% (RT)
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Supelco
Thioglycolic acid solution, ~80% in H2O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Supelco
Ampicillin, analytical standard
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium hydrosulfite, technical grade
SAFC
Isopropyl β-D-1-thiogalactopyranoside
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Thioglycolic acid, 98%
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)