  • Search Results
  • Engineering the genome of Thermus thermophilus using a counterselectable marker.
Engineering the genome of Thermus thermophilus using a counterselectable marker.
Journal of bacteriology (2015-01-22)
Jennifer F Carr, Michael E Danziger, Athena L Huang, Albert E Dahlberg, Steven T Gregory
ABSTRACT
Thermus thermophilus is an extremely thermophilic bacterium that is widely used as a model thermophile, in large part due to its amenability to genetic manipulation. Here we describe a system for the introduction of genomic point mutations or deletions using a counterselectable marker consisting of a conditionally lethal mutant allele of pheS encoding the phenylalanyl-tRNA synthetase α-subunit. Mutant PheS with an A294G amino acid substitution renders cells sensitive to the phenylalanine analog p-chlorophenylalanine. Insertion of the mutant pheS allele via a linked kanamycin resistance gene into a chromosomal locus provides a gene replacement intermediate that can be removed by homologous recombination using p-chlorophenylalanine as a counterselective agent. This selection is suitable for the sequential introduction of multiple mutations to produce a final strain unmarked by an antibiotic resistance gene. We demonstrated the utility of this method by constructing strains bearing either a point mutation in or a precise deletion of the rrsB gene encoding 16S rRNA. We also used this selection to identify spontaneous, large-scale deletions in the pTT27 megaplasmid, apparently mediated by either of the T. thermophilus insertion elements ISTth7 and ISTth8. One such deletion removed 121 kb, including 118 genes, or over half of pTT27, including multiple sugar hydrolase genes, and facilitated the development of a plasmid-encoded reporter system based on β-galactosidase. The ability to introduce mutations ranging from single base substitutions to large-scale deletions provides a potentially powerful tool for engineering the genome of T. thermophilus and possibly other thermophiles as well. Thermus thermophilus is an extreme thermophile that has played an important part in the development of both biotechnology and basic biological research. Its suitability as a genetic model system is established by its natural competence for transformation, but the scarcity of genetic tools limits the kinds of manipulations that can currently be performed. We have developed a counterselectable marker that allows the introduction of unmarked deletions and point mutations into the T. thermophilus genome. We find that this marker can also be used to select large chromosomal deletions apparently resulting from aberrant transposition of endogenous insertion sequences. This system has the potential to advance the genetic manipulation of this important model organism.
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥98% (TLC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
4-Nitrophenyl β-D-galacto­pyran­oside, ≥98% (enzymatic)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥99.0% (TLC)
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material