  • Search Results
  • Top2a identifies and provides epigenetic rationale for novel combination therapeutic strategies for aggressive prostate cancer.
Top2a identifies and provides epigenetic rationale for novel combination therapeutic strategies for aggressive prostate cancer.
Oncotarget (2015-01-22)
Jason S Kirk, Kevin Schaarschuch, Zafardjan Dalimov, Elena Lasorsa, ShengYu Ku, Swathi Ramakrishnan, Qiang Hu, Gissou Azabdaftari, Jianmin Wang, Roberto Pili, Leigh Ellis
ABSTRACT
Progression of aggressive prostate cancers (PCa) with androgen receptor splice variants or neuroendrocrine features is currently untreatable in the clinic. Therefore novel therapies are urgently required. We conducted RNA-seq using tumors from a unique murine transplant mouse model which spontaneously progresses to metastatic disease. Differential gene expression analysis revealed a significant increase of topoisomerase IIα, Top2a (Top2a) in metastatic tumors. Interrogation of human data revealed that increased Top2a expression in primary tumors selected patients with more aggressive disease. Further, significant positive correlation was observed between Top2a and the histone methyltransferase, Ezh2. Combination of the Top2 poison etoposide with the Ezh2 inhibitor GSK126 or DZNep significantly increased cell death in vitro in murine and human prostate cancer cell lines. Additionally, combination therapy extended time to progression and increased therapeutic efficacy in vivo. Overall, our studies demonstrate that patients screened for Top2a and Ezh2 expression would exhibit significant response to a combinational treatment involving low dose etoposide combined with Ezh2 inhibition. In addition, our data suggests that this combination therapeutic strategy is beneficial against aggressive PCa, and provides strong rationale for continued clinical development.
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Etoposide for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
3,3′-Diaminobenzidine, ≥97% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
3,3′-Diaminobenzidine, 98%
Sigma-Aldrich
R1881, ≥98% (HPLC)
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Etoposide, European Pharmacopoeia (EP) Reference Standard