  • Search Results
  • Susceptibility breakpoints and target values for therapeutic drug monitoring of voriconazole and Aspergillus fumigatus in an in vitro pharmacokinetic/pharmacodynamic model.
Susceptibility breakpoints and target values for therapeutic drug monitoring of voriconazole and Aspergillus fumigatus in an in vitro pharmacokinetic/pharmacodynamic model.
The Journal of antimicrobial chemotherapy (2014-02-20)
Maria Siopi, Eleftheria Mavridou, Johan W Mouton, Paul E Verweij, Loukia Zerva, Joseph Meletiadis
ABSTRACT
Although voriconazole reached the bedside 10 years ago and became the standard care in the treatment of invasive aspergillosis, reliable clinical breakpoints are still in high demand. Moreover, this has increased due to the recent emergence of azole resistance. Four clinical wild-type and non-wild-type A. fumigatus isolates with voriconazole CLSI MICs in the range of 0.125-2 mg/L were tested in an in vitro pharmacokinetic (PK)/pharmacodynamic (PD) model. Mouse PK was simulated and in vitro data were compared with in vivo outcome. Human PK was simulated and susceptibility breakpoints and trough levels required for optimal treatment were determined for the CLSI and EUCAST methods after 48 h and the gradient concentration MIC test strip (MTS) method after 24 h using the in vitro PK/PD relationship and Monte Carlo simulation. The in vitro PK/PD target (95% CI) associated with 50% of the maximal antifungal activity (EC50) was 28.61 (16.18-50.61), close to the in vivo EC50 of 14.67 (9.31-21.58) fAUC0-24/CLSI MIC. When human PK was simulated, the EC50 was 24.7 (17.9-35.6) fAUC0-12/CLSI MIC and it was associated with 6 week survival in clinical studies of invasive pulmonary aspergillosis. Target attainment rates were ≤5% (0%-24%), 42% (16%-58%), 68% (54%-75%) and ≥79% (73%-86%) for isolates with CLSI MICs ≥2, 1, 0.5 and ≤0.25 mg/L, respectively. A trough/CLSI MIC ratio of 2 was required for optimal treatment. The susceptible/intermediate/resistant breakpoints were determined to be 0.25/0.5-1/2 mg/L for CLSI, 0.5/1-2/4 mg/L for EUCAST and 0.25/0.375-1/1.5 mg/L for MTS. These susceptibility breakpoints and target values for therapeutic drug monitoring could be used to optimize voriconazole therapy against A. fumigatus.
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Chloramphenicol, γ-irradiated
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Voriconazole, ≥98% (HPLC)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)