  • Search Results
  • Frequent disruption of chromodomain helicase DNA-binding protein 8 (CHD8) and functionally associated chromatin regulators in prostate cancer.
Frequent disruption of chromodomain helicase DNA-binding protein 8 (CHD8) and functionally associated chromatin regulators in prostate cancer.
Neoplasia (New York, N.Y.) (2014-12-17)
Nathan A Damaschke, Bing Yang, Michael L Blute, Chee Paul Lin, Wei Huang, David F Jarrard
ABSTRACT
Abnormal expression and function of chromatin regulators results in the altered chromatin structure seen in cancer. The chromatin regulator CTCF, its cofactor CHD8, and antagonistic paralogue BORIS have wide-ranging effects on gene regulation. Their concurrent expression and regulation was examined in benign, localized, and metastatic prostate cancer (PCa) arrays with extended follow-up using an automated quantitative imaging system, VECTRA. Epithelial staining was quantified and compared against a range of clinicopathologic variables. CHD8 expression was decreased in HGPIN, localized, and metastatic PCa compared to benign (P < .001). CHD8 promoter hypermethylation, assessed by Quantitative Pyrosequencing, occurred in over 45% of primary cancers in this population as well as the TGCA database. Treatment of cell lines with the demethylating agent 5-Aza-2'-deoxycytidine reinduced expression. An interesting dichotomy for CHD8 was observed within primary cancers, with higher nuclear protein expression associated with adverse clinical outcomes including extracapsular extension (P = .007), presence of metastases (P = .025) and worse PSA-recurrence free survival (P = .048). CHD8 outperformed Gleason score and predicted biochemical failure within intermediate grade prostate cancers. The BORIS/CTCF expression ratio increased in localized (P = .03) and metastatic PCa (P = .006) and was associated with higher Gleason score (P = .02), increased tumor volume (P = .02) and positive margins (P = .04). Per cell heterogeneity of expression revealed all protein expression to be more heterogeneous in cancerous tissue (both P < .001), especially high grade (P < .01). In the first detailed analysis in cancer, a marked loss of CHD8 expression and increased BORIS/CTCF ratio indicate frequent disruption of CTCF and its effector genes in PCa.
Sigma-Aldrich
Trimesic acid, 95%
Sigma-Aldrich
Trimethylaluminum, 97%
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in toluene
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in hexanes
Sigma-Aldrich
Trimethylaluminum, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminum solution, 2.0 M in heptane