  • Search Results
  • Comparation sensory characteristic, non-volatile compounds, volatile compounds and antioxidant activity of MRPs by novel gradient temperature-elevating and traditional isothermal methods.
Comparation sensory characteristic, non-volatile compounds, volatile compounds and antioxidant activity of MRPs by novel gradient temperature-elevating and traditional isothermal methods.
Journal of food science and technology (2015-02-20)
Meigui Huang, Xiaoming Zhang, Eric Karangwa
ABSTRACT
A novel gradient temperature-elevating Maillard reaction was developed to effectively produce light-colored MRPs. The main purpose of the present study was to compare the color, taste characteristic, non-volatile compounds, volatile compounds and antioxidant activity of MRPs prepared by the novel gradient temperature-elevating and traditional isothermal methods. The product prepared from soybean peptide, D-xylose and L-cysteine by gradient temperature-elevating method was termed MRP-60. Isothermal method was used to produce light and dark colored MRP, called PXC and PX, respectively. MRP-60 has not only the lowest brown intensity (A420, 0.088), also the highest overall acceptability taste from sensory evaluation. MRP-60 showed the lowest bitterness which could be attributed to the lowest content of bitter amino acids (1.48 mg/ml); MRP-60 also showed indistinctive mouthfulness compared with PXC which elucidated by the percentage of compounds with a molecular weight of 1,000-5,000 Da (15.03 %) and high antioxidant activity. Then, the MRPs prepared by gradient temperature-elevating method successfully attain desirable taste and high antioxidant activity.
Product Description
Xylose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
SAFC
L-Cysteine
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)