MilliporeSigma
  • miR-124 negatively regulates osteogenic differentiation and in vivo bone formation of mesenchymal stem cells.
miR-124 negatively regulates osteogenic differentiation and in vivo bone formation of mesenchymal stem cells.
Journal of cellular biochemistry (2014-11-27)
Abdul S Qadir, Soyoun Um, Heesu Lee, Kyunghwa Baek, Byoung Moo Seo, Gene Lee, Gwan-Shik Kim, Kyung Mi Woo, Hyun-Mo Ryoo, Jeong-Hwa Baek
ABSTRACT
MicroRNAs are novel key regulators of cellular differentiation. Dlx transcription factors play an important role in osteoblast differentiation, and Dlx5 and Dlx2 are known targets of miR-124. Therefore, in the present study, we investigated the regulatory effects of miR-124 on the osteogenic differentiation and in vivo bone formation of mesenchymal stem cells (MSCs). During osteogenic induction by BMP2, the expression levels of miR-124 were inversely correlated with those of osteogenic differentiation marker genes in human and mouse bone marrow-derived MSCs, MC3T3-E1 cells and C2C12 cells. The overexpression of a miR-124 mimic significantly decreased the expression levels of Dlx5, Dlx3, and Dlx2, whereas the silencing of miR-124 with hairpin inhibitors significantly increased the expression of these Dlx genes. Luciferase reporter assays demonstrated that miR-124 directly targets the 3'UTRs of Dlx3, Dlx5, and Dlx2. The overexpression of a miR-124 mimic suppressed the osteogenic marker gene expression levels, alkaline phosphatase activity and matrix mineralization, which were all significantly increased by the overexpression of a miR-124 inhibitor. When ectopic bone formation was induced by the subcutaneous transplantation of human bone marrow-derived MSCs in nude mice, MSCs overexpressing a miR-124 inhibitor significantly enhanced woven bone formation compared with control MSCs. However, MSCs overexpressing a miR-124 mimic exhibited increased adipocyte differentiation at the expense of ectopic bone formation. These results suggest that miR-124 is a negative regulator of osteogenic differentiation and in vivo bone formation and that the targeting of Dlx5, Dlx3, and Dlx2 genes partly contributes to this inhibitory effect exerted by miR-124.
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard