  • Search Results
  • Protective effect of N-acetylcysteine against DNA damage and S-phase arrest induced by ochratoxin A in human embryonic kidney cells (HEK-293).
Protective effect of N-acetylcysteine against DNA damage and S-phase arrest induced by ochratoxin A in human embryonic kidney cells (HEK-293).
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2014-05-07)
Qian Yang, Lei Shi, Kunlun Huang, Wentao Xu
ABSTRACT
N-acetylcysteine (NAC) has recently gained particular interest as a beneficial antioxidant. This study investigated the protective effects of NAC against ochratoxin A (OTA)-induced DNA damage and S-phase arrest in human embryonic kidney cells (HEK-293). OTA exposure results in nephrotoxicity, hepatotoxicity as well as immunotoxicity; and, in the present study, the toxicity of OTA toward HEK-293 cells was explored by analyzing the involvement of the oxidative pathway. It was found that OTA treatment led to oxidative damage; meanwhile, OTA treatment induced significant DNA damage and S-phase arrest by down-regulating cyclin A2, cyclin E1, and CDK2 expression. However, NAC pretreatment alleviated OTA-induced ROS overproduction, the loss of mitochondrial membrane potential (ΔΨm), and the decrease in superoxide dismutase (SOD) activity. NAC pretreatment was also discovered to attenuate OTA-induced DNA damage using the comet assay and by determining the expression of γ-H2AX. In addition, NAC pretreatment partly ameliorated OTA-induced S-phase arrest by preventing the down-regulation of cyclin A2, cyclin E1 and CDK2 expression in HEK-293 cells. All of these results demonstrated that oxidative damage was involved in OTA-induced DNA damage and cell cycle arrest in HEK-293 cells. Therefore, NAC has the potential to reverse the DNA damage and S-phase arrest induced by OTA.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture