MilliporeSigma
Labeling graphene oxygen groups with europium.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-11-19)
Adriano Ambrosi, Martin Pumera
ABSTRACT
Graphene-related materials contain chemically bonded oxygen atoms in the form of epoxy, hydroxy, carboxy, and carbonyl groups. It is important to determine the quantity of oxygen atoms and to understand their position on the graphene sheet. However, visualization of these groups by standard methods is a challenge. Here, we utilize europium(III) as a selective label for oxygen-containing groups. We studied three different graphene-related materials: 1) graphene oxide, 2) chemically reduced graphene oxide, and 3) thermally reduced graphene oxide (the number of oxygen containing groups decreases from material 1 to 3). We show that it is possible to efficiently use Eu as a label of oxygen-containing groups. This Eu label could be applied to determine the precise location of oxygen-containing groups on graphene sheets and also induce novel optical, electrochemical, and catalytic properties.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Sulfuric acid, 99.999%
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
N,N-Dimethylformamide, analytical standard
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%