  • Search Results
  • Trace analysis of selected hormones and sterols in river sediments by liquid chromatography-atmospheric pressure chemical ionization-tandem mass spectrometry.
Trace analysis of selected hormones and sterols in river sediments by liquid chromatography-atmospheric pressure chemical ionization-tandem mass spectrometry.
Journal of chromatography. A (2014-09-04)
Ivana Matić, Svetlana Grujić, Zorica Jauković, Mila Laušević
ABSTRACT
In this paper, development and optimization of new LC-MS method for determination of twenty selected hormones, human/animal and plant sterols in river sediments were described. Sediment samples were prepared using ultrasonic extraction and clean up with silica gel/anhydrous sodium sulphate cartridge. Extracts were analyzed by liquid chromatography-linear ion trap-tandem mass spectrometry, with atmospheric pressure chemical ionization. The optimized extraction parameters were extraction solvent (methanol), weight of the sediment (2 g) and time of ultrasonic extraction (3× 10 min). Successful chromatographic separation of hormones (estriol and estrone, 17α- and 17β-estradiol) and four human/animal sterols (epicoprostanol, coprostanol, α-cholestanol and β-cholestanol) that have identical fragmentation reactions was achieved. The developed and optimized method provided high recoveries (73-118%), low limits of detection (0.8-18 ng g(-1)) and quantification (2.5-60 ng g(-1)) with the RSDs generally lower than 20%. Applicability of the developed method was confirmed by analysis of six river sediment samples. A widespread occurrence of human/animal and plant sterols was found. The only detected hormone was mestranol in just one sediment sample.
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture