  • Search Results
  • Induction of mesenchymal stem cell differentiation and cartilage formation by cross-linker-free collagen microspheres.
Induction of mesenchymal stem cell differentiation and cartilage formation by cross-linker-free collagen microspheres.
European cells & materials (2014-09-03)
M Mathieu, S Vigier, M N Labour, C Jorgensen, E Belamie, D Noël
ABSTRACT
Because of poor self-healing ability, joint cartilage can undergo irreversible degradation in the course of various diseases or after injury. A promising approach for cartilage engineering consists of using of mesenchymal stem cells (MSC) and a differentiation factor combined with an injectable carrier biomaterial. We describe here a novel synthesis route for native collagen microspheres that does not involve the use of potentially toxic crosslinking agents. An emulsion was formed between a type I collagen solution and perfluorinated oil, stabilised by a biocompatible triblock perfluorinated copolymer surfactant. Spherical microparticles of fibrillar collagen were formed through a sol-gel transition induced by ammonia vapours. Electron microscopy observations showed that these self-cross-linked microspheres were constituted by a gel of striated collagen fibrils. Microspheres that were loaded with transforming growth factor beta (TGF-β)3 progressively released this differentiation factor over a four weeks period. Human MSC rapidly adhered to TGF-β3-loaded microspheres and, after 21 d of culture, exhibited typical chondrocyte morphology and produced an uncalcified matrix made of the predominant cartilage components, aggrecan and type II collagen, but devoid of the hypertrophic marker type X collagen. Subcutaneous co-injection of MSC and TGF-β3-loaded microspheres in mice consistently led to the formation of a cartilage-like tissue, which was however hypertrophic, calcified and vascularised. In conclusion, we developed cross-linker free collagen microspheres that allowed chondrogenic differentiation of MSC in vitro and in vivo.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-Methylmorpholine, purified by redistillation, ≥99.5%
Sigma-Aldrich
4-Methylmorpholine, ReagentPlus®, 99%
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
4-Methylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
4-Methylmorpholine, purum, ≥98.0% (GC)
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Anti-Aggrecan Antibody, Chemicon®, from rabbit
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use