  • Search Results
  • Microbiota and metabolites of aged bottled gueuze beers converge to the same composition.
Microbiota and metabolites of aged bottled gueuze beers converge to the same composition.
Food microbiology (2015-01-15)
Freek Spitaels, Simon Van Kerrebroeck, Anneleen D Wieme, Isabel Snauwaert, Maarten Aerts, Anita Van Landschoot, Luc De Vuyst, Peter Vandamme
ABSTRACT
Gueuze beers are prepared by mixing young and old lambic beers and are bottle-refermented spontaneously for aging. The present study analyzed the microbiota and metabolites present in gueuze beers that were aged between a few months and up to 17 years. Yeasts were cultivated from all beers sampled, but bacteria could not be grown from beers older than 5 years. Lactic acid and ethyl lactate concentrations increased steadily during aging, whereas ethanol concentrations remained constant. The concentrations of isoamyl acetate and ethyl decanoate decreased during the aging process. Hence, ethyl lactate and ethyl decanoate can be considered as positive and negative gueuze beer-aging metabolite biomarkers, respectively. Nevertheless, considerable bottle-to-bottle variation in the metabolite profiles was found, which hindered the generalization of the effects seen during the aging of the gueuze beers examined, but which illustrated the unique character of the lambic beers. The present results further indicate that gueuze beers are preferably aged for less than 10 years.
Sigma-Aldrich
Propionic acid, BioReagent, suitable for insect cell culture, ~99%
Sigma-Aldrich
Maltotriose, ≥90% (HPLC)
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Propionic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Isobutyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)
Supelco
Propionic acid, analytical standard
Sigma-Aldrich
Propionic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Isobutyric acid, puriss. p.a., ≥99.5%
Sigma-Aldrich
Propionic acid, natural, 99%, FG
Sigma-Aldrich
Isobutyric acid, 99%
Sigma-Aldrich
Maltohexaose, ≥65% (HPLC)
Supelco
Isobutyric acid, analytical standard
Sigma-Aldrich
Isobutyric acid, ≥99%, FCC, FG
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propionic acid, ≥99.5%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Divinylbenzene, technical grade, 80%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG