  • Search Results
  • Aptamer-conjugated theranostic hybrid graphene oxide with highly selective biosensing and combined therapy capability.
Aptamer-conjugated theranostic hybrid graphene oxide with highly selective biosensing and combined therapy capability.
Faraday discussions (2014-10-04)
Bhanu Priya Viraka Nellore, Avijit Pramanik, Suhash Reddy Chavva, Sudarson Sekhar Sinha, Christen Robinson, Zhen Fan, Rajashekhar Kanchanapally, Justin Grennell, Ivy Weaver, Ashton T Hamme, Paresh Chandra Ray
ABSTRACT
Cancer is a life-threatening disease, which is rapidly becoming a global pandemic. Driven by this need, here we report for the first time an aptamer-conjugated theranostic magnetic hybrid graphene oxide-based assay for highly sensitive tumor cell detection from blood samples with combined therapy capability. AGE-aptamer-conjugated theranostic magnetic nanoparticle-attached hybrid graphene oxide was developed for highly selective detection of tumor cells from infected blood samples. Experimental data indicate that hybrid graphene can be used as a multicolor luminescence platform for selective imaging of G361 human malignant melanoma cancer cells. The reported results have also shown that indocyanine green (ICG)-bound AGE-aptamer-attached hybrid graphene oxide is capable of combined synergistic photothermal and photodynamic treatment of cancer. Targeted combined therapeutic treatment using 785 nm near-infrared (NIR) light indicates that the multimodal therapeutic treatment is highly effective for malignant melanoma cancer therapy. The reported data show that this aptamer-conjugated theranostic graphene oxide-based assay has exciting potential for improving cancer diagnosis and treatment.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Sodium borohydride, hydrogen-storage grade, 98%
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether