  • Search Results
  • Erythromycin-induced genotoxicity and hepatotoxicity in mice pups treated during prenatal and postnatal period.
Erythromycin-induced genotoxicity and hepatotoxicity in mice pups treated during prenatal and postnatal period.
Fundamental & clinical pharmacology (2013-10-15)
Preeti Singh, Lubhan Singh, Sambhu Charan Mondal, Sokindra Kumar, Ishwari Narayan Singh
ABSTRACT
The main aim of this study was to investigate the oxidative stress, genotoxicity and cytotoxicity of erythromycin (EMC) in pups of treated dams in gestation as well as the lactation period (LP). The two doses of EMC were compared using intraperitoneal (i.p.) route in two different periods, that is, in gestation period and in the LP. The rationale behind selection of i.p. route is because of the fact that EMC gets degrades in acidic pH of the stomach. The doses of EMC used were clinically equivalent dose (CED; EMC 14.2 mg/kg, i.p.) and a lower dose (EMC 10 mg/kg, i.p.) than CED. EMC toxicities in mice pups were evaluated using various parameters such as micronucleus (MN) test in peripheral blood and bone marrow, malondialdehyde (MDA) assay, glutathione (GSH reduced) assay and histopathological assessment in liver tissue. The CED of EMC led to a significant increase in MDA and decreased in GSH concentration in pups' liver tissue in both gestation and LPs and also to a significant increase in MN frequency in both peripheral blood and bone marrow cells of pups. There were no significant toxicities at a lower dose than CED. There were also some chronic findings with liver histopathological examination at CED. It is thus concluded that EMC accentuates the oxidative stress, cytotoxicity and DNA damage in pups of their postnatal life; hence, EMC should be avoided in the pregnancy and also in the LP.
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, for molecular biology
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, natural, 97%, FG
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, for molecular biology