  • Search Results
  • Evaluation of the cytotoxic effects of PLGA coated iron oxide nanoparticles as a carrier of 5- fluorouracil and mega-voltage X-ray radiation in DU145 prostate cancer cell line.
Evaluation of the cytotoxic effects of PLGA coated iron oxide nanoparticles as a carrier of 5- fluorouracil and mega-voltage X-ray radiation in DU145 prostate cancer cell line.
IEEE transactions on nanobioscience (2014-07-23)
Zahra Hajikarimi, Samideh Khoei, Sepideh Khoee, Seied Rabi Mahdavi
ABSTRACT
The purpose of this study was to investigate the uptake and cytotoxic effects of magnetic poly lactic-co-glycolic acid (PLGA)-coated iron oxide nanoparticles as a carrier of 5-fluorouracil (5-FU) and X-ray on the level of proliferation capacity of DU145 prostate carcinoma cell line in monolayer culture. Following monolayer culture, DU 145 cells were treated with different concentrations of 5-FU or 5-FU loaded nanoparticles for 24 h and 2Gy X-ray (6 Mega-voltage (MV)). The rate of nanoparticles penetration was then measured using atomic adsorption spectroscopy (AAS). The cytotoxicity effect of these nanoparticles with/ without X-ray radiation was evaluated using colony formation assay. Spectroscopy results showed that iron content and therefore the cellular uptake of 5-FU loaded nanoparticles increased with increasing nanoparticle concentrations. Further, the proliferation capacity of the cells decreased with the increase of 5-FU and 5- FU loaded nanoparticle concentrations in combination with X-ray radiation. However the extent of reduction in colony number following treatment with 5-FU-loaded nanoparticles in combination with 2Gy of megavoltage X-ray radiation was significantly more than for free 5-FU. Thus, drug-loaded nanoparticles could deliver 5-FU more efficiently into the cells. PLGA coated iron oxide nanoparticles are therefore effective drug delivery vehicles for 5-FU. PLGA coated iron oxide nanoparticles are biocompatible and this coating is an appropriate surface that can penetrate into the cells.
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Nitric acid, puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%
Sigma-Aldrich
Nitric acid, red, fuming, HNO3 >90 %
Sigma-Aldrich
Nitric acid, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Nitric acid, ACS reagent, ≥90.0%
Supelco
Nitric acid concentrate, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Nitric acid, puriss. p.a., 65.0-67.0%
Sigma-Aldrich
Nitric acid, puriss. p.a., ≥65% (T)
Sigma-Aldrich
Nitric acid, ACS reagent, 70%
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Supelco
5-Fluorouracil, analytical standard
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N