  • Search Results
  • Analysis of MUC4 expression in human pancreatic cancer xenografts in immunodeficient mice.
Analysis of MUC4 expression in human pancreatic cancer xenografts in immunodeficient mice.
Anticancer research (2014-07-31)
Daniel Ansari, Monika P Bauden, Agata Sasor, Chinmay Gundewar, Roland Andersson
ABSTRACT
Mucin 4 (MUC4) is a cell surface glycoprotein that is overexpressed in most pancreatic tumors. The aim of the present study was to characterize MUC4 expression in experimental pancreatic cancer in order to clarify the correlation between MUC4 and pancreatic cancer histology in vivo. Pancreatic xenograft tumors were generated in immunodeficient mice (n=15) by subcutaneous injection of MUC4(+) human pancreatic cancer cell lines Capan-1, HPAF-II or CD18/HPAF. MUC4 immunoreactivity was compared between the cancer models. Alpha-smooth muscle actin (α-SMA) was used to identify cancer-associated fibroblasts and the amount of collagen fibers was quantified with sirius red. Tumor incidence was 100%. Tumor size showed no difference across groups (p=0.796). The median MUC4 count was highest in Capan-1 tumors (p=0.002). α-SMA and collagen extent were also highest in Capan-1 tumors (p=0.018). The Capan-1 xenograft model could serve as a valuable resource to test new therapeutic strategies targeting MUC4 in pancreatic cancer.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)