  • Search Results
  • Hiwi facilitates chemoresistance as a cancer stem cell marker in cervical cancer.
Hiwi facilitates chemoresistance as a cancer stem cell marker in cervical cancer.
Oncology reports (2014-08-15)
Wei Liu, Qing Gao, Kunlun Chen, Xiang Xue, Mu Li, Qian Chen, Gaixia Zhu, Ya Gao
ABSTRACT
Hiwi, also named PiwiL1, is a human homologue of the Piwi family which is associated with stem cells and is overexpressed in several types of cancers. In the present study, we aimed to investigate the role of Hiwi in cervical carcinogenesis. Immunochemical analysis showed a significantly higher frequency of Hiwi staining in high-grade squamous intraepithelial lesions (HSILs) and cervical cancer tissues when comparing with the frequency in normal cervices. Particularly, Hiwi staining was restricted to basal cells of the normal cervix and was associated with the progression of cervical cancer and chemotherapy resistance. We further found that ectopic Hiwi increased the chemical resistance in SiHa cells, and silencing of Hiwi in HeLa cells decreased the cell viability. In addition, as a cancer stem cell marker, Hiwi promoted the tumorsphere formation in vitro and tumorigenicity in vivo and elevated the expression of several stem cell self-renewal-associated transcription factors, in spite of inhibited the proliferation. These results suggest that Hiwi may participate in the carcinogenesis of cervical cancer and may be a potential therapeutic target molecule for cervical cancers.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard