  • Search Results
  • The effect of chronic stress on anti-angiogenesis of sunitinib in colorectal cancer models.
The effect of chronic stress on anti-angiogenesis of sunitinib in colorectal cancer models.
Psychoneuroendocrinology (2014-12-02)
Jie Liu, Guo-Hua Deng, Jie Zhang, Ying Wang, Xiang-Yu Xia, Xin-Mei Luo, Yao-Tiao Deng, Sha-Sha He, Yin-Yan Mao, Xing-Chen Peng, Yu-Quan Wei, Yu Jiang
ABSTRACT
Epidemiological and experimental evidence has shown that psychological stress can propel cancer progression. However, its role in anti-angiogenic therapy is not well understood. We previously found that exogenous norepinephrine attenuated the effect of sunitinib, a multi-targeted anti-angiogenic agent, in a mouse melanoma model. Here, we further evaluated the effects of chronic stress on sunitinib therapy in colorectal cancer models. We found that chronic restraint stress markedly weakened the efficacy of sunitinib, primarily through promoting the expression of vascular endothelial growth factor (VEGF) and interleukin-8 (IL-8) to stimulate tumor angiogenesis in vivo. This effect could be sufficiently mimicked by exogenous norepinephrine and blocked by the β-antagonist propranolol. In vitro, norepinephrine up-regulated expression of VEGF and IL-8 in sunitinib-treated cancer cells mainly through the β-adrenoceptor-cAMP-PKA signaling pathway. Norepinephrine also abrogated sunitinib-induced inhibition of cancer cell migration, but had no effect on direct anti-proliferative activity of sunitinib on cancer cells. These findings suggest that psychological stress might attenuate anti-angiogenic therapy primarily through activating beta-adrenergic signaling to promote tumor angiogenesis. It is also suggested that β-blockers might improve anti-angiogenic outcome under psychological stress.
Product Description
USP
Propranolol hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Supelco
(±)-Propranolol hydrochloride, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard