  • Search Results
  • Comparison of centrally injected tryptophan-related substances inducing sedation in acute isolation stress-induced neonatal chicks.
Comparison of centrally injected tryptophan-related substances inducing sedation in acute isolation stress-induced neonatal chicks.
Pharmacology, biochemistry, and behavior (2014-12-03)
Junki Yoshida, Edi Erwan, Vishwajit Sur Chowdhury, Yumi Ogino, Asako Shigemura, D Michael Denbow, Mitsuhiro Furuse
ABSTRACT
In the present study, we first focused on the function of l-tryptophan (TRP) metabolites which are synthesized in different metabolic pathways, namely, the kynurenine (KYN) pathway and serotonin (5-HT) pathway during an acute isolation stress. When l-TRP metabolites were intracerebroventricularly injected on an equimolar basis (100 nmol), 5-HT induced a sedative effect in neonatal chicks. Additionally, plasma corticosterone, dopamine, 5-HT, and its metabolite 5-hydroxyindoleacetic acid concentrations were increased in the diencephalon of the 5-HT treated group compared with other groups. Second, the two doses (400 or 800 nmol) of l- and d-TRP were compared under a corticotrophin-releasing hormone-augmented social isolation stress. When comparing the efficacy between l- and d-TRP against stress behavior, both amino acids had a similar effect and quickly suppressed distress vocalizations. Finally, d-amino acid levels in the diencephalon and telencephalon were measured but d-TRP was not found. These results indicate that l- and d-TRP induce the same effect in attenuating stress but the mode of action of TRP derivatives, namely 5-HT differs during an acute isolation stress in neonatal chick. The absence of d-TRP in the diencephalon further suggests that instead of being an endogenous factor it may play role as a pharmacological factor.
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
Chloromethane, ≥99.5%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Supelco
Corticosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Chloromethane solution, 1.0 M in tert-butyl methyl ether, anhydrous
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Chloromethane solution, 1.0 M in diethyl ether
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Supelco
Chloromethane solution, 200 μg/mL in methanol, analytical standard