  • Search Results
  • Low concentrations of trichosanthin induce apoptosis and cell cycle arrest via c-Jun N-terminal protein kinase/mitogen-activated protein kinase activation.
Low concentrations of trichosanthin induce apoptosis and cell cycle arrest via c-Jun N-terminal protein kinase/mitogen-activated protein kinase activation.
Molecular medicine reports (2014-10-30)
Duo Zhang, Bin Chen, Jian Zhou, Lin Zhou, Qing Li, Fei Liu, Kuang-Yen Chou, Lei Tao, Li-Ming Lu
ABSTRACT
Trichosanthin (TCS) is a type I ribosome--inactivating protein, which inhibits cell viability in human epithelial type 2 (HEp-2) and AMC-HN-8 human laryngeal epidermoid carcinoma cells. Although TCS is a potential chemotherapeutic agent, its mechanism of action remains to be elucidated. In the present study, HEp-2 and AMC-HN-8 cells were treated with different concentrations of TCS combined with or without cisplatin. After 5 days of successive treatment, different experimental groups were detected using a cell counting kit-8 and the collected supernatants were analyzed using a lactate dehydrogenase kit. Flow cytometric assays were performed to detect apoptosis and cell cycle arrest in the HEp-2 and AMC-HN-8 cells, reverse transcription quantitative polymerase chain reaction was performed to detect the levels of p27, p21WAF and western blot analysis was performed to detect changes in c-Jun N-terminal protein kinase (JNK)/phosphorylated (phospho)-JNK, p38/phospho-p38, extracellular signal-regulated kinase (ERK)/phospho-ERK, caspase-3 and caspase-9 in the HEp-2 and AMC-HN-8 cancer cells. TCS significantly inhibited the cell viability of the HEp-2 and AMC-HN-8 cells, independently of necrosis. TCS induced apoptosis and increased the percentage of HEp-2 and AMC-HN-8 cells in the S-phase of the cell cycle. In addition, the JNK/mitogen-activated protein kinase (MAPK) pathway was activated by TCS in the HEp-2 and AMC-HN-8 cells. Low concentrations of TCS also induced apoptosis and S-phase cell cycle arrest in the HEp-2 and AMC-HN-8 cells. The antitumor effects of TCS may be associated with JNK/MAPK activation.
Product Description
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Cisplatin impurity A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard