  • Search Results
  • A tunable Caco-2/HT29-MTX co-culture model mimicking variable permeabilities of the human intestine obtained by an original seeding procedure.
A tunable Caco-2/HT29-MTX co-culture model mimicking variable permeabilities of the human intestine obtained by an original seeding procedure.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-04-08)
Arnaud Béduneau, Camille Tempesta, Stéphane Fimbel, Yann Pellequer, Vincent Jannin, Frédéric Demarne, Alf Lamprecht
ABSTRACT
Standard monoculture models utilizing Caco-2 monolayers were extensively used to mimic the permeability of the human intestinal barrier. However, they exhibit numerous limitations such as the lack of mucus layer, an overestimation of the P-gp-mediated efflux and a low paracellular permeability. Here, we suggest a new procedure to set up an in vitro model of intestinal barrier to adjust gradually the properties of the absorption barrier. Mucin-secreting HT29-MTX cells were added to Caco-2 absorptive cells in a Transwell® at different time intervals. Effects of seeding day of HT29-MTX on the paracellular permeability of lucifer yellow (LY) and on the P-gp-mediated efflux of rhodamine 123 were investigated. Apparent permeability of the rhodamine 123 in the secretory direction was highly dependent on the seeding day of goblet cells. Transepithelial electrical resistance values and LY transport across the co-cultures in the apical-to-basolateral direction were intermediary between single Caco-2 and HT29-MTX models. Early seeding days of HT29-MTX allowed increasing the fraction of goblet cells in the co-culture. Co-culture permeability was unchanged between 21 and 30 days after Caco-2 seeding, corresponding to the period of use for Caco-2-based cell models. Thus, the HT29-MTX seeding day was a key factor to set up an in vitro intestinal model with tailor-made barrier properties in terms of P-gp expression and paracellular permeability.
Product Description
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Dextrose, meets EP, BP, JP, USP testing specifications, anhydrous
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Dextrose, Pharmaceutical Secondary Standard; Certified Reference Material