MilliporeSigma
  • Propionibacterium acnes activates the NLRP3 inflammasome in human sebocytes.
Propionibacterium acnes activates the NLRP3 inflammasome in human sebocytes.
The Journal of investigative dermatology (2014-05-14)
Zheng Jun Li, Dae Kyoung Choi, Kyung Cheol Sohn, Min Seok Seo, Hae Eul Lee, Young Lee, Young Joon Seo, Young Ho Lee, Ge Shi, Christos C Zouboulis, Chang Deok Kim, Jeung Hoon Lee, Myung Im
ABSTRACT
Propionibacterium acne and sebaceous glands are considered to have an important role in the development of acne. Although information regarding the activation of innate immunity by P. acnes in the sebaceous gland is limited, different P. acnes phylotypes and a higher prevalence of follicular P. acnes macrocolonies/biofilms in sebaceous follicles of skin biopsies from acne compared with control skin and occasionally single P. acnes clusters in single sebaceous glands have been detected. In this study, we investigated whether P. acnes activates the inflammasome in human sebaceous glands in vivo and in vitro. We found that IL-1β expression was upregulated in sebaceous glands of acne lesions. After stimulation of human sebocytes with P. acnes, the activation of caspase-1 and secretion of IL-1β were enhanced significantly. Moreover, knocking down the expression of NLRP3 abolished P. acnes-induced IL-1β production in sebocytes. The activation of the NLRP3 inflammasome by P. acnes was dependent on protease activity and reactive oxygen species generation. Finally, we found that NALP3-deficient mice display an impaired inflammatory response to P. acnes. These results suggest that human sebocytes are important immunocompetent cells that induce the NLRP3 inflammasome, and that P. acnes-induced IL-1β activation in sebaceous glands may have a role in combating skin infections and in acne pathogenesis.
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis