  • Search Results
  • Development and analytical validation of an enzyme-linked immunosorbent assay for the measurement of feline tumor necrosis factor α in serum.
Development and analytical validation of an enzyme-linked immunosorbent assay for the measurement of feline tumor necrosis factor α in serum.
Veterinary clinical pathology (2014-07-01)
Joerg M Steiner, Panagiotis G Xenoulis, Viktoria M Schwierk, Jan S Suchodolski
ABSTRACT
The role of tumor necrosis factor alpha (TNF-α), a cytokine shown to play a crucial role in human Crohn's disease patients, has not been documented in cats with chronic enteropathies. Also, currently, no validated assay for measurement of TNF-α in cats is available. The objective of this study was to develop and analytically validate an enzyme-linked immunosorbent assay (ELISA) for the quantification of TNF-α in serum from cats. A sandwich ELISA was developed and analytically validated by assessment of detection limit, linearity, accuracy, precision, and reproducibility. A control range for serum fTNF-α concentration in healthy cats was established. In addition, serum concentrations of fTNF-α in 39 cats with chronic enteropathies were compared with those in 20 healthy cats. The detection limit of the assay was 38.4 ng/L. Observed-to-expected ratios for serial dilutions of 4 serum samples ranged from 75.1% to 111.9%. Observed-to-expected ratios for spiking recovery for 4 serum samples ranged from 91.3% to 129.7%. Coefficients of variation for intra- and inter-assay variability ranged from 3.9% to 7.6% and from 7.8% to 12.5%, respectively. The control range of the assay was < 38.4-223.5 ng/L. Serum concentrations of feline TNF-α were significantly higher in cats with chronic enteropathies and diarrhea than in cats with chronic enteropathies without diarrhea, or in healthy control cats. The ELISA described here was suitable for the quantification of fTNF-α in feline serum and should facilitate research into the importance of TNF-α in cats with chronic enteropathies.
Product Description
Supelco
Mettler-Toledo Calibration substance ME 18555, Benzoic acid, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bovine TNFα / Tumor Necrosis Factor alpha ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Mouse Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Human Tumor Necrosis Factor α ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Supelco
Benzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Melting point standard 121-123°C, analytical standard
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Supelco
Benzoic acid, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Rat Tumor Necrosis Factor α ELISA Kit, for cell and tissue lysates
USP
Benzoic acid, United States Pharmacopeia (USP) Reference Standard
Benzoic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
SS Benzoic Acid, 2000 μg/mL in dichloromethane, analytical standard
Supelco
Benzoic acid, Standard for quantitative NMR, TraceCERT®
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder