  • Search Results
  • Dexamethasone suppresses neurosteroid biosynthesis via downregulation of steroidogenic enzyme gene expression in human glioma GI-1 cells.
Dexamethasone suppresses neurosteroid biosynthesis via downregulation of steroidogenic enzyme gene expression in human glioma GI-1 cells.
Biological & pharmaceutical bulletin (2014-07-06)
Fuyuko Koibuchi, Natsumi Ritoh, Ryohei Aoyagi, Megumi Funakoshi-Tago, Hiroomi Tamura
ABSTRACT
Emerging evidence indicates that stress hormone glucocorticoids (GC) are an important modulator of brain development and function. To investigate whether GCs modulate neurosteroid biosynthesis in neural cells, we studied the effects of GCs on steroidogenic gene expression in human glioma GI-1 cells. The GC dexamethasone (Dex) reduced steroidogenic acute regulatory protein (StAR), CYP11A1 and 3β-hydroxysteroid dehydrogenase gene expression in a dose- and GC receptor-dependent manner. In addition to its effects on steroidogenic gene expression, Dex also reduced de novo synthesis of progesterone (PROG). Furthermore, Dex inhibited all-trans retinoic acid (ATRA) and vitamin D3-induced steroidogenic gene expression and PROG production. This suggests that GC regulates steroidogenic gene expression in neural cells via cross-talk with the two fat-soluble vitamins, A and D. The relationship between the effects of GCs on neurosteroid biosynthesis and on cognitive behaviors and hippocampal neural activity is also discussed herein.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Vitamin D3 solution, 100 μg/mL in ethanol, 97% (CP)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Cholecalciferol (D3), analytical standard
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
5-Pregnen-3β-ol-20-one, ≥98%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Tretinoin, European Pharmacopoeia (EP) Reference Standard
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Progesterone, meets USP testing specifications
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.