  • Search Results
  • Controlled iontophoretic delivery of pramipexole: electrotransport kinetics in vitro and in vivo.
Controlled iontophoretic delivery of pramipexole: electrotransport kinetics in vitro and in vivo.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-02-15)
Dhaval R Kalaria, Pratikkumar Patel, Virginia Merino, Vandana B Patravale, Yogeshvar N Kalia
ABSTRACT
The objective of the study was to investigate the anodal iontophoretic delivery of pramipexole (PRAM), a dopamine agonist used for the treatment of Parkinson's disease, in order to determine whether therapeutic amounts of the drug could be delivered across the skin. Preliminary iontophoretic experiments were performed in vitro using porcine ear and human abdominal skin. These were followed by a pharmacokinetic study in male Wistar rats to determine the drug input rate in vivo. Stability studies revealed that after current application (0.5 mA/cm(2) for 6h), the solution concentration of PRAM was only 60.2 ± 5.3% of its initial value. However, inclusion of sodium metabisulfite (0.5%), an antioxidant, increased this to 97.2 ± 3.1%. Iontophoretic transport of PRAM across porcine skin in vitro was studied as a function of current density (0.15, 0.3, 0.5 mA/cm(2)) and concentration (10, 20, 40 mM). Increasing the current density from 0.15 to 0.3 and 0.5 mA/cm(2), resulted in 2.5- and 4-fold increases in cumulative permeation, from 309.5 ± 80.2 to 748.8 ± 148.1 and 1229.1 ± 138.6 μg/cm(2), respectively. Increasing the PRAM concentration in solution from 10 to 20 and 40 mM resulted in a 2-fold increase in cumulative permeation (816.4 ± 123.3, 1229.1 ± 138.6 and 1643.6 ± 201.3 μg/cm(2), respectively). Good linearity was observed between PRAM flux and both the applied current density (r(2)=0.98) and drug concentration in the formulation (r(2)=0.99). Co-iontophoresis of acetaminophen showed that electromigration was the dominant electrotransport mechanism (accounting for >80% of delivery) and that there was no inhibition of electroosmotic flow at any current density. Cumulative iontophoretic permeation across human and porcine skin (after 6h at 0.5 mA/cm(2)) was also shown to be statistically equivalent (1229.1 ± 138.6 and 1184.8 ± 236.4 μg/cm(2), respectively). High transport and delivery efficiencies were achieved for PRAM (up to 7% and 58%, respectively). The plasma concentration profiles obtained in the iontophoretic studies in vivo (20 mM PRAM; 0.5 mA/cm(2) for 5h) were modelled using constant and time-variant input models; the latter gave a superior quality fit. The drug input rate in vivo suggested that PRAM electrotransport rates would be sufficient for therapeutic delivery and the management of Parkinsonism.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, tested according to Ph. Eur., anhydrous
Supelco
Citric acid, analytical standard
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Sodium metabisulfite, ACS reagent, ≥97.0%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, 99.93%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Supelco
Methanol, analytical standard
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis