  • Search Results
  • In vitro antifungal susceptibility of Scopulariopsis brevicaulis isolates.
In vitro antifungal susceptibility of Scopulariopsis brevicaulis isolates.
Medical mycology (2014-07-23)
Magdalena Skóra, Anna B Macura, Małgorzata Bulanda
ABSTRACT
In humans, Scopulariopsis is mainly associated with onychomycoses, rarely with cutaneous infections or with invasive mycoses. However, during the last two decades, deep infections caused by members of this genus have been increasing. Scopulariopsis brevicaulis is the most common species described as an etiologic agent of human disease. Previous antifungal susceptibility studies indicate that this species is resistant in vitro to the broad-spectrum antifungal agents that are available today. Here, we describe the antifungal activity of amphotericin B, terbinafine, ciclopirox, itraconazole, ketoconazole, and voriconazole against 40 S. brevicaulis isolates. Antifungal susceptibility tests were performed using a modified Clinical and Laboratory Standards Institute M38-A2 procedure. The results showed that itraconazole had the highest minimal inhibitory concentration (MIC) of >16 mg/l; amphotericin B, voriconazole, and ketoconazole MICs were ranging from 4 to >16 mg/l, 8 to >16 mg/l, and 8 to >16 mg/l, respectively; and the best activity was found with terbinafine and ciclopirox with MICs ranging from 0.5 to 16 mg/l and 1 to 8 mg/l, respectively.
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
SAFC
MOPS
USP
Sodium bicarbonate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Glutamine