  • Search Results
  • Oxidative stress in the brain of cigarette smoke-induced noxiousness: neuroprotective role of Sesbania grandiflora.
Oxidative stress in the brain of cigarette smoke-induced noxiousness: neuroprotective role of Sesbania grandiflora.
Metabolic brain disease (2014-09-14)
Thiyagarajan Ramesh, Chandrabose Sureka, Shanmugham Bhuvana, Vavamohaideen Hazeena Begum
ABSTRACT
Cigarette smoking is concerned as a major risk factor in the development of various neurological disorders. Oxidative stress is suggested as a possible contributing factor in the pathogenesis of cigarette smoking-induced toxicity. Therefore, the present study was intended to evaluate the neuroprotective role of Sesbania grandiflora (S. grandiflora) against chronic cigarette smoke induced oxidative damage in rat brain. Adult male Wistar-Kyoto rats were exposed to cigarette smoke for a period of 90 days and consecutively treated with S. grandiflora aqueous suspension (SGAS, 1,000 mg/kg body weight per day by oral gavage) for a period of 3 weeks. Lipid peroxidation and antioxidants status were analyzed in the brain. Rats exposed to cigarette smoke showed significant increase in conjugated diens (CD), hydroperoxides (HP) and malendialdehyde (MDA) levels with concomitant decrease in superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPx), glutathione reductase (GR), glutathione-S-transferase (GST) and glucose-6-phosphate dehydrogenase (G6PDH) activities and the levels of reduced glutathione (GSH), vitamin C and vitamin E. Also cigarette smoke-exposure resulted in a marked increase in copper and decrease in zinc, manganese and selenium levels in brain. Administration of SGAS attenuates lipid peroxidation, enhanced the antioxidant status, restored the levels of micronutrients and retained the brain histology. The results of our study indicate that chronic cigarette smoke-exposure accelerates oxidative stress, thereby disquieting the brain defensive mechanism and S. grandiflora protects the brain from the oxidative damage through its biopotency.
Product Description
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Glutathione, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate sodium salt, pkg of 5 mg (per vial)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material