  • Search Results
  • Applicability of a newly developed bioassay for determining bioactivity of anti-inflammatory compounds in release studies--celecoxib and triamcinolone acetonide released from novel PLGA-based microspheres.
Applicability of a newly developed bioassay for determining bioactivity of anti-inflammatory compounds in release studies--celecoxib and triamcinolone acetonide released from novel PLGA-based microspheres.
Pharmaceutical research (2014-08-29)
Hsiao-yin Yang, Maarten van Dijk, Ruud Licht, Michiel Beekhuizen, Mattie van Rijen, Martina Källrot Janstål, F Cumhur Öner, Wouter J A Dhert, Detlef Schumann, Laura B Creemers
ABSTRACT
To develop a bio-assay for measuring long-term bioactivity of released anti-inflammatory compounds and to test the bioactivity of celecoxib (CXB) and triamcinolone acetonide (TA) released from a new PLGA-based microsphere platform. Human osteoarthritic chondrocytes were plated according to standardized procedures after batch-wise harvest and cultured for 3 days to prevent cell confluency and changes in cell behaviour. Prostaglandin E2 (PGE2) production stimulated by TNFα was used as a parameter of inflammation. A novel microsphere platform based on PTE-functionalised PLGA was used to incorporate CXB and TA. Loaded microspheres were added to transwells overlying the cells, with transfer of the wells to new cell cultures every 3 days. Inhibition of PGE2 production was determined over a period of 21 days. PLGA(75:25)-PTE microspheres were prepared and loaded with CXB and TA at 86 and 97% loading efficiency, respectively. In the bioactivity assay, PGE2 levels induced by TNFα were reduced to an average of 30% using microspheres loaded with 0.1 nmol CXB per transwell; with microspheres loaded with 0.1 nmol TA, PGE2 production was initially reduced to 3% and gradually recovered to 30% reduction. At 1 nmol loading, PGE2 was inhibited to 0-7% for CXB-loaded microspheres, and 0-28% for TA-loaded microspheres. We present a novel sustained release bioactivity assay which provides an essential link between in vitro buffer-based release kinetics and in vivo application. Novel PLGA-based microspheres loaded with TA and CXB showed efficient anti-inflammatory effects over time.
Sigma-Aldrich
Glycolide, ≥99%
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Supelco
Dichloromethane solution, contains 10 % (v/v) methanol
Supelco
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
Supelco
Dichloromethane, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Supelco
Dichloromethane, analytical standard, stabilized
Supelco
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
Supelco
Methanol solution, contains 0.1 % (v/v) trifluoroacetic acid, 5 % (v/v) water, suitable for HPLC
Sigma-Aldrich
(3S)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione, 98%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer