  • Search Results
  • Mycobacterium tuberculosis 38-kDa antigen induces endoplasmic reticulum stress-mediated apoptosis via toll-like receptor 2/4.
Mycobacterium tuberculosis 38-kDa antigen induces endoplasmic reticulum stress-mediated apoptosis via toll-like receptor 2/4.
Apoptosis : an international journal on programmed cell death (2014-12-30)
Yun-Ji Lim, Ji-Ae Choi, Jeong-Hwan Lee, Chul Hee Choi, Hwa-Jung Kim, Chang-Hwa Song
ABSTRACT
Endoplasmic reticulum (ER) stress responses play critical roles in the pathogenesis of tuberculosis. To investigate the regulatory role of the ER stress response in 38-kDa antigen-induced apoptosis, we examined the relationship between the ER stress response and apoptosis in bone marrow-derived macrophages (BMDMs) stimulated with Mycobacterium tuberculosis antigen (38-kDa Ag). The expression of ER molecular chaperones, including C/EBP homologous protein (CHOP), glucose-regulated protein (Bip) and phosphorylated alpha subunit of eukaryotic initiation factor 2, was induced in BMDMs stimulated with the 38-kDa Ag. Interestingly, 38-kDa Ag-stimulation induced apoptosis via activation of caspase-12, -9 and -3. However, 38-kDa Ag-induced apoptosis was significantly reduced in TLR2- and TLR4-deficient macrophages. Because toll-like receptors (TLRs) initiate the activation of mitogen-activated protein kinase (MAPK) signaling cascades, we evaluated the effect of MAPK activation on ER stress. The 38-kDa Ag activated Jun N-terminal kinase, extracellular signal-regulated kinase and p38 phosphorylation. MAPK signaling induced the secretion of proinflammatory cytokines such as MCP-1, TNF-α and IL-6. The 38-kDa Ag-induced MCP-1 was especially associated with the induction of MCP-1-induced protein (MCPIP), which increased the generation of reactive oxygen species (ROS) and ER stress. To investigate the role of MCPIP in ROS-induced ER stress by 38-kDa Ag stimulation, we transfected MCPIP siRNA into RAW264.7 cells before 38-kDa Ag stimulation, and measured the generation of ROS and expression of ER molecular chaperones. ROS production and CHOP expression were decreased by the silencing of MCPIP induction. Our results demonstrate that the expression of MCPIP by 38-kDa Ag stimulation is increased through a TLR-MAPK-dependent signaling pathway, and leads to ER stress-induced apoptosis. In conclusion, MCPIP is important for host defense mechanisms in mycobacterial pathogenesis.
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Sigma-Aldrich
N-Acetyl-L-cysteine, BioXtra, ≥99% (TLC)
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Supelco
N-Acetyl-L-cysteine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Acetylcysteine, European Pharmacopoeia (EP) Reference Standard
USP
Acetylcysteine, United States Pharmacopeia (USP) Reference Standard