  • Search Results
  • Toxoplasma gondii nucleus coding apicoplast protein ACP synthesis and trafficking in delayed death.
Toxoplasma gondii nucleus coding apicoplast protein ACP synthesis and trafficking in delayed death.
Parasitology research (2015-01-08)
Liang Wu, Jin Shen, Yupei Zhou, Xiao Wang, Lamei Wu, Xugan Jiang, Shengxia Chen
ABSTRACT
This study aimed to explore Toxoplasma gondii nucleus coding apicoplast protein acyl carrier protein (ACP) synthesis and trafficking in delayed death. The recombinant T. gondii ACP was expressed by prokaryotic expression method, and anti-ACP polyclonal antibody was obtained from rabbit immune. T. gondii "delayed death" was induced by clindamycin (CLDM), and ACP transcription was determined by real-time PCR assay. The expression of ACP with transit type (t-ACP) and mature type (m-ACP) was determined by Western blotting with anti-ACP polyclonal antibody. The mutant-expressed ACP fused with green fluorescent protein (GFP) tag was constructed by pHX-ACP-GFP. The distribution of ACP in "delayed death" was observed by ACP-GFP fusion protein with a confocal microscope. T. gondii ACP transcription and t-ACP expression had no significant decrease in the early 4 h of "delayed death," but there has been a significant decrease in 6 h. The expression of m-ACP had a significant decrease in 4 h which occurred earlier than the t-ACP expression. The number of brightly dot green fluorescence in ACP-GFP mutant decreased with prolonged time. There was very little brightly dot green fluorescence in ACP-GFP mutant when treated with CLDM for 6 h. CLDM could suppress apicoplast proliferation and induce T. gondii "delayed death"; however, it could not directly suppress nucleus coding ACP transcription and expression. T. gondii lacking of apicoplast had a barrier of transit peptide cleavage and t-ACP could not be transformed into m-ACP. The reason for the decrease in ACP expression could be due to excessive t-ACP synthesis in tachyzoites resulting in a negative feedback for the ACP coding gene transcription.
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Xanthine, ≥99.5% (HPLC), purified by recrystallization
Sigma-Aldrich
Xanthine, BioUltra, ≥99%
Sigma-Aldrich
Xanthine, ≥99%
Sigma-Aldrich
Potassium hydroxide on alumina
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethanol, anhydrous, denatured
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, meets USP testing specifications
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, HPLC/spectrophotometric grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC