  • Search Results
  • Application of magnetic solid phase extraction for separation and determination of aflatoxins B ₁ and B₂ in cereal products by high performance liquid chromatography-fluorescence detection.
Application of magnetic solid phase extraction for separation and determination of aflatoxins B ₁ and B₂ in cereal products by high performance liquid chromatography-fluorescence detection.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-05-13)
Mahdi Hashemi, Zohreh Taherimaslak, Somayeh Rashidi
ABSTRACT
A simple and sensitive method based on the magnetic solid phase extraction with modified magnetic nanoparticles followed by high performance liquid chromatography with fluorescence detection has been developed for extraction and determination of aflatoxins B1 (AFB1) and B2 (AFB2) in cereal products. Magnetic nanoparticle coated with 3-(trimethoxysilyl)-1-propanthiol (TMSPT) and modified with 2-amino-5-mercapto-1,3,4-thiadiazole (AMT) was used as an antibody-free adsorbent. Under the optimal conditions, the calibration curves for AFB1 and AFB2 were linear in the ranges of 0.2-15 μg L(-1) and 0.04-3 μg L(-1), respectively. Detection limit was 0.041 μg L(-1) for AFB1 and 0.013 μg L(-1) for AFB2. The proposed method was successfully applied to the determination of AFB1 and AFB2 in spiked corn and rice samples with an average recovery of 93.5%. The results demonstrated that the developed method is simple, rapid, inexpensive, accurate and remarkably free from interference effects.
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
SAFC
Sodium chloride solution, 5 M