  • Search Results
  • A series of new lanthanoid thioarsenates: insights into the influence of lanthanide contraction on the formation of new lanthanoid thioarsenates.
A series of new lanthanoid thioarsenates: insights into the influence of lanthanide contraction on the formation of new lanthanoid thioarsenates.
Dalton transactions (Cambridge, England : 2003) (2015-03-20)
Jian Zhou, Rong-Qing Zhao, Tao Yang, Xing Liu, Hong-Ping Xiao, Hua-Hong Zou, Xiao-Feng Tan
ABSTRACT
A series of new lanthanoid thioarsenates [Ln(teta)(μ-η(1):η(2):η(1)-As(III)S3)]n {Ln = Ce (Ia), Pr (Ib), Nd (Ic), and Sm (Id); teta = triethylenetetramine} and [Ln(teta)(en)(μ-η(1):η(1):η(1)-As(V)S4)]n {Ln = La (IIa), Ce (IIb), Pr (IIc), and Nd (IId); en = ethylenediamine} were prepared by the solvothermal reaction of K3AsO3, S, LnCl3 and organic amines and structurally characterized. Compounds Ia–d crystallise in the orthorhombic space group Aba2 and display 1-D neutral chains [Ln(teta)(μ-η(1):η(2):η(1)-As(III)S3)]n, which represent the first examples of 1-D organic hybrid lanthanoid sulfides built up from trigonal-pyramidal [As(III)S3](3-) acting as tetradentate bridging ligands to interlink [Ln(teta)](3+) ions, while compounds IIa–d crystallise in the orthorhombic space group P2(1)2(1)2(1) and consist of other 1-D neutral chains [Ln(teta)(en)(μ-η(1):η(1):η(1)-As(V)S4)]n, which are built up from the linkages of the tetrahedral [As(V)S4](3-) ion and the [Ln(teta)(en)](3+) ion. To learn more about the influence of lanthanide contraction on the formation of lanthanoid thioarsenates, three organic hybrid lanthanoid thioarsenates [Ln(teta)(en)As(V)S4] [Ln = Dy (IIIa), Ho (IIIb), and Tm (IIIc)] with the neutral molecular structure type in the monoclinic centrosymmetric space group P2(1)/c are also presented. Their optical and magnetic properties have been investigated, and density functional theory calculations of Ia and IIa have also been performed.
Sigma-Aldrich
Mercury, electronic grade, 99.9999% trace metals basis
Sigma-Aldrich
Mercury, ≥99.99% trace metals basis
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis