  • Search Results
  • A non-pediocin low molecular weight antimicrobial peptide produced by Pediococcus pentosaceus strain IE-3 shows increased activity under reducing environment.
A non-pediocin low molecular weight antimicrobial peptide produced by Pediococcus pentosaceus strain IE-3 shows increased activity under reducing environment.
BMC microbiology (2014-08-28)
Pradip K Singh, Shalley Sharma, Annu Kumari, Suresh Korpole
ABSTRACT
Species of the genus Pediococcus are known to produce antimicrobial peptides such as pediocin-like bacteriocins that contain YGNGVXC as a conserved motif at their N-terminus. Until now, the molecular weight of various bacteriocins produced by different strains of the genus Pediococcus have been found to vary between 2.7 to 4.6 kD. In the present study, we characterized an antimicrobial peptide produced by P. pentosaceus strain IE-3. Antimicrobial peptide was isolated and purified from the supernatant of P. pentosaceus strain IE-3 grown for 48 h using cation exchange chromatography and reversed-phase high-performance liquid chromatography (RP-HPLC) techniques. While MALDI-TOF MS experiments determined the precise molecular mass of the peptide to be 1701.00 Da, the de novo sequence (APVPFSCTRGCLTHLV) of the peptide revealed no similarity with reported pediocins and did not contain the YGNGVXC conserved motif. Unlike pediocin-like bacteriocins, the low molecular weight peptide (LMW) showed resistance to different proteases. Moreover, peptide treated with reducing agent like dithiothreitol (DTT) exhibited increased activity against both Gram-positive and Gram-negative test strains in comparison to native peptide. However, peptide treated with oxidizing agent such as hydrogen peroxide (H2O2) did not show any antimicrobial activity. To our knowledge this is the lowest molecular weight peptide produced by members of the genus Pediococcus. The low molecular weight peptide shared amino acid arrangement with N-terminal sequence of Class IIa, pediocin-like bacteriocins and showed increased activity under reducing conditions. Antimicrobial peptides active under reduced conditions are valuable for the preservation of processed foods like meat, dairy and canned foods where low redox potential prevails.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)