  • Search Results
  • Protein kinase C in the wood frog, Rana sylvatica: reassessing the tissue-specific regulation of PKC isozymes during freezing.
Protein kinase C in the wood frog, Rana sylvatica: reassessing the tissue-specific regulation of PKC isozymes during freezing.
PeerJ (2014-09-12)
Christopher A Dieni, Kenneth B Storey
ABSTRACT
The wood frog, Rana sylvatica, survives whole-body freezing and thawing each winter. The extensive adaptations required at the biochemical level are facilitated by alterations to signaling pathways, including the insulin/Akt and AMPK pathways. Past studies investigating changing tissue-specific patterns of the second messenger IP3 in adapted frogs have suggested important roles for protein kinase C (PKC) in response to stress. In addition to their dependence on second messengers, phosphorylation of three PKC sites by upstream kinases (most notably PDK1) is needed for full PKC activation, according to widely-accepted models. The present study uses phospho-specific immunoblotting to investigate phosphorylation states of PKC-as they relate to distinct tissues, PKC isozymes, and phosphorylation sites-in control and frozen frogs. In contrast to past studies where second messengers of PKC increased during the freezing process, phosphorylation of PKC tended to generally decline in most tissues of frozen frogs. All PKC isozymes and specific phosphorylation sites detected by immunoblotting decreased in phosphorylation levels in hind leg skeletal muscle and hearts of frozen frogs. Most PKC isozymes and specific phosphorylation sites detected in livers and kidneys also declined; the only exceptions were the levels of isozymes/phosphorylation sites detected by the phospho-PKCα/βII (Thr638/641) antibody, which remained unchanged from control to frozen frogs. Changes in brains of frozen frogs were unique; no decreases were observed in the phosphorylation levels of any of the PKC isozymes and/or specific phosphorylation sites detected by immunoblotting. Rather, increases were observed for the levels of isozymes/phosphorylation sites detected by the phospho-PKCα/βII (Thr638/641), phospho-PKCδ (Thr505), and phospho-PKCθ (Thr538) antibodies; all other isozymes/phosphorylation sites detected in brain remained unchanged from control to frozen frogs. The results of this study indicate a potential important role for PKC in cerebral protection during wood frog freezing. Our findings also call for a reassessment of the previously-inferred importance of PKC in other tissues, particularly in liver; a more thorough investigation is required to determine whether PKC activity in this physiological situation is indeed dependent on phosphorylation, or whether it deviates from the generally-accepted model and can be "overridden" by exceedingly high levels of second messengers, as has been demonstrated with certain PKC isozymes (e.g., PKCδ).
Product Description
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Supelco
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Acrylamide, certified reference material, TraceCERT®
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)