MilliporeSigma
  • Matrix stiffening and β1 integrin drive subtype-specific fibroblast accumulation in lung cancer.
Matrix stiffening and β1 integrin drive subtype-specific fibroblast accumulation in lung cancer.
Molecular cancer research : MCR (2014-10-05)
Marta Puig, Roberto Lugo, Marta Gabasa, Alícia Giménez, Adriana Velásquez, Roland Galgoczy, Josep Ramírez, Abel Gómez-Caro, Óscar Busnadiego, Fernando Rodríguez-Pascual, Pere Gascón, Noemí Reguart, Jordi Alcaraz
ABSTRACT
The crucial role of tumor-associated fibroblasts (TAF) in cancer progression is now clear in non-small cell lung cancer (NSCLC). However, therapies against TAFs are limited due to a lack of understanding in the subtype-specific mechanisms underlying their accumulation. Here, the mechanical (i.e., matrix rigidity) and soluble mitogenic cues that drive the accumulation of TAFs from major NSCLC subtypes: adenocarcinoma (ADC) and squamous cell carcinoma (SCC) were dissected. Fibroblasts were cultured on substrata engineered to exhibit normal- or tumor-like stiffnesses at different serum concentrations, and critical regulatory processes were elucidated. In control fibroblasts from nonmalignant tissue, matrix stiffening alone increased fibroblast accumulation, and this mechanical effect was dominant or comparable with that of soluble growth factors up to 0.5% serum. The stimulatory cues of matrix rigidity were driven by β1 integrin mechano-sensing through FAK (pY397), and were associated with a posttranscriptionally driven rise in β1 integrin expression. The latter mechano-regulatory circuit was also observed in TAFs but in a subtype-specific fashion, because SCC-TAFs exhibited higher FAK (pY397), β1 expression, and ERK1/2 (pT202/Y204) than ADC-TAFs. Moreover, matrix stiffening induced a larger TAF accumulation in SCC-TAFs (>50%) compared with ADC-TAFs (10%-20%). In contrast, SCC-TAFs were largely serum desensitized, whereas ADC-TAFs responded to high serum concentration only. These findings provide the first evidence of subtype-specific regulation of NSCLC-TAF accumulation. Furthermore, these data support that therapies aiming to restore normal lung elasticity and/or β1 integrin-dependent mechano regulation may be effective against SCC-TAFs, whereas inhibiting stromal growth factor signaling may be effective against ADC-TAFs. This study reveals distinct mechanisms underlying the abnormal accumulation of tumor-supporting fibroblasts in two major subtypes of lung cancer, which will assist the development of personalized therapies against these cells.
Product Description
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Selenium, pellets, < 5mm, ≥99.999%
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Supelco
Picric acid solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)