  • Search Results
  • Activation of IL6/IGFIR confers poor prognosis of HBV-related hepatocellular carcinoma through induction of OCT4/NANOG expression.
Activation of IL6/IGFIR confers poor prognosis of HBV-related hepatocellular carcinoma through induction of OCT4/NANOG expression.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-01-08)
Te-Sheng Chang, Yu-Chih Wu, Ching-Chi Chi, Wei-Chi Su, Pey-Jium Chang, Kam-Fai Lee, Tao-Hsin Tung, Jui Wang, Jun-Jen Liu, Shui-Yi Tung, Liang-Mou Kuo, Hong-Nerng Ho, Thai-Yen Ling, Yen-Hua Huang
ABSTRACT
To unravel the role of interleukin (IL)-6 and insulin-like growth factor (IGF)-I receptor (IGFIR) in expressing stemness-related properties and to evaluate the prognostic values of pluripotent transcription factor OCT4/NANOG, and IGFIR in hepatocellular carcinoma (HCC). Serum levels of IL6 were detected using ELISA assays (n = 120). The effects of IL6/IGFI on stemness expression in HCC were examined using OCT4/NANOG promoter luciferase reporter, RNA interference, secondary sphere formation, side population, and xenograft animal models. The OCT4/NANOG protein and phospho-IGFI receptor (p-IGFIR) in tissues were detected by Western blotting (n = 8) and immunohistochemical staining (n = 85). OCT4, NANOG, and IGFIR expression levels in tissues (n = 191) were analyzed by real-time qRT-PCR and was correlated with early tumor recurrence using the Kaplan-Meier survival analysis. A high positive correlation between the expression levels of OCT4/NANOG and IGFIR/p-IGFIR in human HCC tissues was observed. The concurrent expression of OCT4/NANOG/IGFIR was mostly confined to hepatitis B virus (HBV)-related HCC (HBV-HCC) and was significantly correlated with early tumor recurrence. High serum levels of IL6 were significantly correlated with high OCT4/NANOG expression. IL6 stimulated an autocrine IGFI/IGFIR expression STAT3 dependently, which stimulated stemness-related properties in both the cell lines and the xenografted mouse tumors. The inhibition of IGFIR activation by either RNA interference or by treatment with the inhibitor picropodophyllin (PPP) significantly suppressed the IL6-induced stemness-related properties both in vitro and in vivo. The expression of pluripotency-related genes is associated with early tumor recurrence and is regulated by IL6-induced IGF/IGFIR activation, particularly in HBV-HCC.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets