  • Search Results
  • Projections from the oral pontine reticular nucleus to the spinal cord of the mouse.
Projections from the oral pontine reticular nucleus to the spinal cord of the mouse.
Neuroscience letters (2014-12-03)
Huazheng Liang, Charles Watson, George Paxinos
ABSTRACT
The present study investigated projections of the mouse oral pontine reticular nucleus (PnO) to the spinal cord by (a) injecting a retrograde tracer fluoro-gold (FG) to the lumbar cord and (b) an anterograde tracer biotinylated dextran amine (BDA) to PnO. We found that PnO projects to the entire spinal cord with an ipsilateral predominance. PnO fibers mainly travel in the ipsilateral ventral funiculus in the entire cord, terminating in laminae 7-10 with a lower density of fibers and boutons in lower segments. A small number of fibers travel in the contralateral ventral funiculus in the cervical cord with a similar terminating pattern to the ipsilateral counterpart. The present study is the first demonstration of PnO fiber terminals in the mouse spinal cord. This pathway might be responsible for muscle atonia during REM sleep, but needs physiological research to confirm this.
Sigma-Aldrich
Sucrose, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5%
Supelco
Meprobamate, analytical standard
Sigma-Aldrich
3,3′-Diaminobenzidine, ≥97% (HPLC)
Sigma-Aldrich
3,3′-Diaminobenzidine, 98%
Sigma-Aldrich
N-(3-Maleimidopropionyl)biocytin, ≥75% (HPLC)
Millipore
Sucrose, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Sucrose, Grade II, suitable for plant cell culture
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
Sucrose, European Pharmacopoeia (EP) Reference Standard
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Supelco
Meprobamate solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®