  • Search Results
  • Nuclear actin aggregation is a hallmark of anti-synthetase syndrome-induced dysimmune myopathy.
Nuclear actin aggregation is a hallmark of anti-synthetase syndrome-induced dysimmune myopathy.
Neurology (2015-03-10)
Werner Stenzel, Corinna Preuße, Yves Allenbach, Debora Pehl, Reimar Junckerstorff, Frank L Heppner, Kay Nolte, Eleonora Aronica, Veronika Kana, Elisabeth Rushing, Udo Schneider, Kristl G Claeys, Olivier Benveniste, Joachim Weis, Hans H Goebel
ABSTRACT
To analyze antisynthetase syndrome-associated myositis by modern myopathologic methods and to define its place in the spectrum of idiopathic inflammatory myopathies (IIMs). Skeletal muscle biopsies from antisynthetase syndrome-associated myositis and other IIMs from different institutions worldwide were analyzed by histopathology, quantitative PCR, and electron microscopy. Myonuclear actin filament inclusions were identified as a unique morphologic hallmark of antisynthetase syndrome-associated myositis. Nuclear actin inclusions were never found in dermatomyositis, polymyositis, sporadic inclusion body myositis, autoimmune necrotizing myopathy associated with signal recognition particle or 3-hydroxy-3-methylglutaryl-coenzyme A reductase autoantibodies, or nonspecific myositis associated with other systemic diseases, harboring myositis-associated autoantibodies, and presenting myofiber necrosis. We show that molecules involved in actin filament formation and actin shuttling mechanisms are altered in antisynthetase syndrome, and may thus be involved in pathologic myonuclear actin aggregation. In addition, we have identified a typical topographic distribution of necrotic myofibers predominantly located at the periphery of muscle fascicles accompanied by inflammation and destruction of the perimysial connective tissue. Antisynthetase syndrome-associated myositis is characterized by distinctive myonuclear actin filament inclusions, including rod formations and a typical necrotizing perimysial myositis. This supports the hypothesis that antisynthetase syndrome-associated myositis is unique and should not be grouped among dermatomyositis, polymyositis, sporadic inclusion body myositis, necrotizing autoimmune myositis, or nonspecific myositis. This study provides Class II evidence that for patients with IIMs, the presence of myonuclear actin filament inclusions accurately identifies patients with antisynthetase syndrome-associated myositis (sensitivity 81%, specificity 100%).
Product Description
Supelco
Residual Solvent - Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC)