  • Search Results
  • In vivo and in vitro toxicity and anti-inflammatory properties of gold nanoparticle bioconjugates to the vascular system.
In vivo and in vitro toxicity and anti-inflammatory properties of gold nanoparticle bioconjugates to the vascular system.
Toxicological sciences : an official journal of the Society of Toxicology (2014-09-28)
Mayara Klimuk Uchiyama, Daiana Kotra Deda, Stephen Fernandes de Paula Rodrigues, Carine Cristiane Drewes, Simone Marques Bolonheis, Pedro Kunihiko Kiyohara, Simone Perche de Toledo, Walter Colli, Koiti Araki, Sandra Helena Poliselli Farsky
ABSTRACT
Gold nanoparticle (AuNP) bioconjugates have been used as therapeutic and diagnostic tools; however, in vivo biocompatibility and cytotoxicity continue to be two fundamental issues. The effect of AuNPs (20 nm) conjugated with antibody [immunoglobulin G (IgG)], albumin, protein A, PEG4000, and citrate (cit) were evaluated in vitro using primary human cells of the vascular system. AuNP bioconjugates did not cause lysis of human erythrocytes, apoptosis or necrosis of human leukocytes, and endothelial cells in vitro, although AuNPs had been internalized and detected in the cytoplasm. Moreover, the influence of AuNPs on rheological parameters, blood and vessel wall characteristics was investigated in vivo by intravital microscopy assay using male Wistar rats mesentery microcirculation as model. Intravenous injection of AuNP-IgG or cit-AuNP did not cause hemorrhage, hemolysis or thrombus formation, instead suppressed the leukocyte adhesion to postcapillary vessel walls, an early stage of an inflammatory process. Furthermore, AuNP-IgG abrogated the expression of platelet-endothelial cell adhesion molecule-1, chemotaxis, and oxidative burst activation on neutrophils after leukotriene B4 stimulation, a membrane receptor-dependent stimulus, thus confirming their anti-inflammatory effects in vitro. The expression of oxidative burst activation was also suppressed after stimulating AuNP-IgG-treated neutrophils with lipid-soluble phorbol myristate acetate (PMA), confirming the direct intracellular action of AuNP-IgG on the inflammatory process in vitro. Our in vitro and in vivo experimental approaches highlighted the great potentiality of AuNP bioconjugates for therapeutic and diagnostic applications by parenteral routes.
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ≥98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Trypan Blue, Dye content 60 %
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, >98%
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥99% (calc. to the dried substance)
Supelco
N-(1-Naphthyl)ethylenediamine dihydrochloride, for determination of sulfonamide and nitrite, ACS reagent, ≥98%
Supelco
Sulfanilamide, VETRANAL®, analytical standard
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Supelco
Sulfanilamide melting point standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Leukotriene B4, ~100 μg/mL in ethanol, ≥97%
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
USP
Sulfanilamide Melting Point Standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
USP
Sulfanilamide, United States Pharmacopeia (USP) Reference Standard
Sulfanilamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Propidium iodide solution, solution (1.0 mg/ml in water)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture