  • Search Results
  • Ligularia fischeri extract protects against oxidative-stress-induced neurotoxicity in mice and PC12 cells.
Ligularia fischeri extract protects against oxidative-stress-induced neurotoxicity in mice and PC12 cells.
Journal of medicinal food (2014-08-19)
Soo Jung Choi, Jae Kyeom Kim, Soo Hwan Suh, Cho Rong Kim, Hye Kyung Kim, Chang-Ju Kim, Gwi Gun Park, Cheung-Seog Park, Dong-Hoon Shin
ABSTRACT
Alzheimer's disease (AD) is pathologically characterized by the presence of amyloid plaques in brain and the overproduction of amyloid beta (Aβ), leading to learning and memory impairment and intense oxidative stress. In this study, the protective effect of Ligularia fischeri extract was investigated using PC12 cells. L. fischeri extract attenuated hydrogen-peroxide-induced DNA fragmentation in cells. In vivo behavioral tests were performed to examine the effects of the extract on amyloid-β peptide1-42-induced impairment of learning and memory in mice. A diet containing the extract increased alternation behaviors in the Y-maze test and step-through latency of passive avoidance task. Moreover, we found that consumption of the extract decreased lipid peroxidation in a biochemical study of brain tissue in mice. High-performance liquid chromatography was used to identify the active compounds in the extract. These results suggest that L. fischeri extract could be protective against Aβ-induced neurotoxicity, possibly due to the antioxidative capacity of its constituent, 3-O-caffeoylquinic acid.
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Phosphoric acid, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O