  • Search Results
  • The role of retinal vessels caliber as a marker of vascular aging in large arteries.
The role of retinal vessels caliber as a marker of vascular aging in large arteries.
Journal of hypertension (2015-04-29)
Luis García-Ortiz, José I Recio-Rodríguez, Cristina Agudo-Conde, Maria C Patino-Alonso, Emiliano Rodríguez-Sánchez, José A Maderuelo-Fernandez, Manuel A Gómez-Marcos
ABSTRACT
To analyze the relationship between retinal vessel caliber and large arterial structure and function in a sample of the general population. A cross-sectional study was conducted among 229 participants from the EVIDENT trial (mean age 55.6±12.1 years, 61.1% women). Retinal photographs were digitized, temporal vessels were measured in an area of 0.5 to 1 disc diameter from the optic disc and the arterio-venous ratio (AVR) was estimated. Vascular structure and function were assessed by evaluating carotid intima-media thickness (IMT), pulse wave velocity (PWV), augmentation index (AIx), ankle brachial index and ambulatory arterial stiffness index. The mean AVR was 0.792 ± 0.082, calculated from an arterial caliber of 109.1 ± 12.5 μm and a venous caliber of 138.7 ± 15.4 μm. A total of 13.1% of the patients presented vascular damage. The AVR was inversely correlated with carotid IMT (r = -0.190, P < 0.01), PWV (r = -0.178, P < 0.01) and AIx (r = -0.187, P < 0.01). In a multiple linear regression analysis, the associations with IMT and with AIx remained after adjusting for possible confounders (β = -0.152, P = 0.026; and β = -18.1, P = 0.037, respectively), but the association with PWV was lost when blood pressure was added (β = -1.520, P = 0.215). In a logistic regression analysis, the odds ratio of the first tertile of mean AVR (<0.76) of having a vascular injury was 7.09 (95% confidence interval 1.78-28.17, P = 0.011) after adjusting for possible confounders. In the general population, lower AVR values are associated with increases in carotid IMT, AIx and PWV. AVR may be a good predictor of asymptomatic vascular damage in large arteries.
Product Description
Supelco
Dibutyl phthalate, Selectophore
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Sigma-Aldrich
Dibutyl phthalate, 99%
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, meets analytical specification of Ph. Eur., BP, E339, 98.5-102.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, puriss. p.a., crystallized, ≥99.0% (T)