  • Search Results
  • Renal tight junction proteins are decreased in cisplatin-induced nephrotoxicity in rats.
Renal tight junction proteins are decreased in cisplatin-induced nephrotoxicity in rats.
Toxicology mechanisms and methods (2014-07-24)
Joyce Trujillo, Eduardo Molina-Jijón, Omar Noel Medina-Campos, Rafael Rodríguez-Muñoz, José Luis Reyes, María L Loredo, Edilia Tapia, Laura Gabriela Sánchez-Lozada, Diana Barrera-Oviedo, José Pedraza-Chaverri
ABSTRACT
Cisplatin (CP) is an antineoplastic agent that induces nephrotoxicity and oxidative stress. It is unknown whether renal tight junction (TJ) proteins expression and localization are modified in CP-induced nephrotoxicity. To study if the expression of the TJ proteins occludin, claudin-2, claudin-5 and zonula occludens-1 (ZO-1) is modified in rats with CP-induced nephrotoxicity. Male Wistar rats (n = 5/group) were injected with saline solution (V group), and the other group (CP group) was injected with a single dose of saline solution and CP (7.5 mg/kg i.p.). Rats were sacrificed 72 h after CP injection and blood, and 24-h urine samples were collected. Several plasma and urinary injury biomarkers as well as renal histopathology lesions, oxidative and nitrosative stress markers were evaluated, and protein levels of ocludin, claudin-2, claudin-5, ZO-1 were measured by Western blot. Statistically significant changes noted with different p < 0.05 versus V. Nephrotoxicity was evident by histological alterations, glycosuria, decrease in creatinine clearance, increase in fractional excretion of sodium, serum creatinine and kidney injury molecule-1. These changes were associated with oxidative/nitrosative stress (increased renal abundance of 3-nitrotyrosine and protein kinase Cβ2 and decreased renal expression of nuclear factor-erythroid-2-related factor 2) and decreased activity of antioxidant enzymes. Finally, it was found that CP-induced renal damage was associated with decreased renal expression of occludin and claudin-2. CP altered the TJ proteins expression and localization in the proximal tubule that was associated with oxidative/nitrosative stress.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 4.00-4.99 %
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 10-15 %
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hypochlorite solution, 6-14% active chlorine basis
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology