MilliporeSigma
  • Trps1 regulates biliary epithelial-mesenchymal transition and has roles during biliary fibrosis in liver grafts: a preliminary study.
Trps1 regulates biliary epithelial-mesenchymal transition and has roles during biliary fibrosis in liver grafts: a preliminary study.
PloS one (2015-04-18)
Cheng Zhe, Fan Yu, Ju Tian, Shuguo Zheng
ABSTRACT
To investigate the role(s) of Trps1 in non-anastomotic biliary stricture (NABS) following liver transplantation. Immunohistochemical and histological techniques were used to detect Trps1, E-cadherin, CK19, vimentin, α-SMA, and collagen deposition. Human intrahepatic biliary epithelial cells (HIBECs) were infected with a Trps1 adenovirus, or transfected with Trps1 short-interfering RNAs (siRNAs). Reverse transcription polymerase chain reaction (RT-PCR) assays and western blotting were used to determine expression levels of epithelial and mesenchymal markers, and Trps1 in HIBECs. Expression of Trps1 and epithelial markers was down-regulated or absent in NABS liver samples. Mesenchymal markers were seen in biliary epithelial cells (BECs), with collagen deposited around the bile duct. Trps1 expression positively correlated with epithelial markers. Expression of epithelial marker mRNAs and proteins in HIBECs decreased with prolonged cold preservation (CP), while mesenchymal marker expression increased. A 12-h CP period led to increased Trps1 mRNA and protein levels. Expression of E-cadherin was increased in HIBECs following Trps1 adenovirus infection and CP/reperfusion injury (CPRI), with vimentin expression levels reduced and CPRI-mediated epithelial-mesenchymal transition (EMT) inhibited. Transfection of HIBECs with Trps1 siRNAs in conjunction with CPRI revealed that E-cadherin expression was decreased, vimentin expression was increased, and CPRI-mediated EMT was promoted. Trps1 is involved in NABS pathogenesis following liver transplantation and negatively correlates with BEC EMT and biliary fibrosis in liver grafts. Trps1 demonstrates antagonistic effects that could reverse EMT.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%