  • Search Results
  • De novo RNA sequencing and transcriptome analysis of Colletotrichum gloeosporioides ES026 reveal genes related to biosynthesis of huperzine A.
De novo RNA sequencing and transcriptome analysis of Colletotrichum gloeosporioides ES026 reveal genes related to biosynthesis of huperzine A.
PloS one (2015-03-24)
Guowei Zhang, Wenjuan Wang, Xiangmei Zhang, Qianqian Xia, Xinmei Zhao, Youngjoon Ahn, Nevin Ahmed, Andreea Cosoveanu, Mo Wang, Jialu Wang, Shaohua Shu
ABSTRACT
Huperzine A is important in the treatment of Alzheimer's disease. There are major challenges for the mass production of huperzine A from plants due to the limited number of huperzine-A-producing plants, as well as the low content of huperzine A in these plants. Various endophytic fungi produce huperzine A. Colletotrichum gloeosporioides ES026 was previously isolated from a huperzine-A-producing plant Huperzia serrata, and this fungus also produces huperzine A. In this study, de novo RNA sequencing of C. gloeosporioides ES026 was carried out with an Illumina HiSeq2000. A total of 4,324,299,051 bp from 50,442,617 high-quality sequence reads of ES026 were obtained. These raw data were assembled into 24,998 unigenes, 40,536,684 residues and 19,790 genes. The majority of the unique sequences were assigned to corresponding putative functions based on BLAST searches of public databases. The molecular functions, biological processes and biochemical pathways of these unique sequences were determined using gene ontology (GO) and Kyoto Encyclopedia of Genes and Genomes (KEGG) assignments. A gene encoding copper amine oxidase (CAO) (unigene 9322) was annotated for the conversion of cadaverine to 5-aminopentanal in the biosynthesis of huperzine A. This gene was also detected in the root, stem and leaf of H. serrata. Furthermore, a close relationship was observed between expression of the CAO gene (unigene 9322) and quantity of crude huperzine A extracted from ES026. Therefore, CAO might be involved in the biosynthesis of huperzine A and it most likely plays a key role in regulating the content of huperzine A in ES026.
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Tropane, 98%
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Supelco
Cadaverine, analytical standard
Supelco
L-Lysine, analytical standard
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%
Sigma-Aldrich
Piperidine, puriss. p.a., ≥99.0% (GC/T)
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Supelco
Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Supelco
Pyridine, analytical standard
Sigma-Aldrich
Piperidine solution, suitable for peptide synthesis, 20% in DMF
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%